This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03368950_cn
See other SLC35E2B CNV Assays ›
Gene Symbol
SLC35E2B
Assay Reference Genome
Location

Chr.1:1671394 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03368950_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728661

Gene Name:

solute carrier family 35 member E2B

Gene Aliases:

SLC35E2

Location:

Chr.1:1661478-1692804 on Build GRCh38

Assay Gene Location:

Within Intron 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC35E2B NM_001110781.2 NP_001104251.1
NM_001290264.1 NP_001277193.1
XM_011542072.1 XP_011540374.1
AB007916.3
AK125619.1
AK307886.1
BC110653.1 AAI10654.1
CR936618.1 CAI56761.1
DA794179.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n54 Chr.1:1663759 - 1696548 on Build GRCh38 Loss SLC35E2B
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion GNB1 CDK11B CDK11A SLC35E2 MMP23A NADK FNDC10 MIB2 SLC35E2B MMP23B
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss SSU72 CDK11B CDK11A SLC35E2 MMP23A FNDC10 MIB2 SLC35E2B MMP23B
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain CDK11B CDK11A MMP23A MIB2 SLC35E2B MMP23B
esv3585010 Chr.1:1670280 - 1751138 on Build GRCh38 Loss CDK11A SLC35E2 MMP23A SLC35E2B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss TNFRSF18 LOC105378949 LINC01128 LOC100132287 LINC00115 FNDC10 LOC100288175 CPSF3L GNB1 LOC284600 MMP23A ACAP3 MIR6726 FAAP20 ATAD3B OR4F16 PLEKHN1 FAM41C PERM1 LOC102725121 CALML6 WASH7P TNFRSF4 LOC100506504 OR4F29 MTND1P23 AGRN MIR6727 MIR6723 NADK LOC105378591 LOC100134822 C1orf159 MMP23B LOC105378948 VWA1 MIR429 LOC100129534 LOC100133331 MORN1 SCNN1D LOC105378592 PRKCZ SSU72 MIR6859-2 MIR200A NOC2L KLHL17 TAS1R3 SAMD11 MIB2 LINC01342 LOC148413 FAM132A ISG15 MIR6859-1 LOC101928626 ANKRD65 SKI LOC100288069 HES4 UBE2J2 LOC100287934 MIR1302-2 FAM87B MRPL20 B3GALT6 RNF223 CCNL2 LOC729737 MIR6808 LOC102724312 LOC107984841 CDK11A LOC105378947 ATAD3A MIR200B TMEM88B CPTP LOC107985729 DDX11L1 LOC107985728 AURKAIP1 SDF4 FAM138A SLC35E2B TMEM52 ATAD3C DVL1 SLC35E2 TTLL10 TTLL10-AS1 OR4F5 LOC105378589 MTND2P28 GABRD TMEM240 CFAP74 LOC100130417 CDK11B PUSL1 MXRA8
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SSU72 GNB1 CDK11B CDK11A SLC35E2 MMP23A NADK FNDC10 MIB2 SLC35E2B MMP23B
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11B CDK11A MMP23A SLC35E2B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss SSU72 CDK11B CDK11A SLC35E2 MMP23A NADK FNDC10 MIB2 SLC35E2B MMP23B
esv3585008 Chr.1:1663929 - 1681002 on Build GRCh38 Loss SLC35E2B
esv3585007 Chr.1:1647187 - 1714507 on Build GRCh38 Loss CDK11B CDK11A MMP23A SLC35E2B
nsv950516 Chr.1:1663141 - 1715961 on Build GRCh38 Deletion CDK11A MMP23A SLC35E2B
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain CDK11B CDK11A SLC35E2 MMP23A NADK SLC35E2B MMP23B
nsv545022 Chr.1:1665581 - 1683909 on Build GRCh38 Loss SLC35E2B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss TNFRSF18 LOC148413 FAM132A LINC01128 LINC00115 FNDC10 LOC100288175 ISG15 CPSF3L ANKRD65 LOC284600 MMP23A ACAP3 LOC100288069 MIR6726 HES4 UBE2J2 LOC100287934 ATAD3B PLEKHN1 FAM41C FAM87B MRPL20 PERM1 B3GALT6 RNF223 CCNL2 TNFRSF4 MIR6808 LOC102724312 AGRN MIR6727 CDK11A ATAD3A MIR200B NADK C1orf159 TMEM88B CPTP LOC107985729 MMP23B LOC105378948 VWA1 MIR429 LOC107985728 AURKAIP1 SDF4 SLC35E2B SCNN1D SSU72 ATAD3C DVL1 SLC35E2 TTLL10 MIR200A TTLL10-AS1 NOC2L KLHL17 TAS1R3 TMEM240 LOC100130417 CDK11B SAMD11 PUSL1 MIB2 MXRA8 LINC01342
nsv545026 Chr.1:1671348 - 1706566 on Build GRCh38 Loss CDK11A MMP23A SLC35E2B
nsv545018 Chr.1:1638499 - 1688102 on Build GRCh38 Loss CDK11B SLC35E2B
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain CDK11B CDK11A SLC35E2 MMP23A SLC35E2B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss LOC148413 LOC105378949 CDK11A ATAD3A NADK FNDC10 LOC105378591 TMEM88B LOC107985729 MMP23B ANKRD65 VWA1 GNB1 MMP23A SLC35E2B PRKCZ SSU72 TMEM52 ATAD3C ATAD3B SLC35E2 LOC105378589 GABRD MRPL20 TMEM240 CFAP74 CDK11B CCNL2 CALML6 MIB2 LOC102724312

Back To Top

More Information


Additional Information:

For this assay, SNP(s) [rs79690428] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0