This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03369061_cn
See other PUSL1 CNV Assays ›
Gene Symbol
PUSL1
Assay Reference Genome
Location

Chr.1:1309127 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03369061_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

126789

Gene Name:

pseudouridylate synthase-like 1

Gene Aliases:

-

Location:

Chr.1:1308562-1311677 on Build GRCh38

Assay Gene Location:

Within Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PUSL1 NM_153339.1 3 207 NP_699170.1
XM_005244720.3 3 259 XP_005244777.1
AK075292.1 3 224 BAC11527.1
AK298218.1 2 117
BC034304.1 3 207 AAH34304.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC100288069 PLEKHN1 CPTP ATAD3A MIR6723 RNF223 LINC00115 MIR429 PUSL1 FAM41C LOC100287934 MIR6727 LOC100129534 LOC107985728 CFAP74 LOC107985729 SKI CDK11A MMP23B B3GALT6 MIR200B GNB1 FAM138A DDX11L1 MIR6808 WASH7P OR4F5 LOC105378949 MMP23A LOC102724312 HES4 LOC105378591 CALML6 SLC35E2 LINC01342 SCNN1D LOC100130417 FAM132A TTLL10-AS1 OR4F29 MRPL20 MIR6859-2 ACAP3 LOC105378592 TNFRSF18 GABRD TNFRSF4 ATAD3B FAAP20 ANKRD65 PERM1 TMEM52 LOC107984841 SLC35E2B ATAD3C NOC2L MIB2 MXRA8 LOC284600 TMEM88B LOC100133331 C1orf159 MTND1P23 LOC148413 CPSF3L LINC01128 ISG15 FNDC10 AURKAIP1 AGRN TAS1R3 OR4F16 SAMD11 LOC100288175 NADK MORN1 VWA1 LOC105378589 CCNL2 FAM87B LOC101928626 SDF4 LOC100132287 TMEM240 LOC100134822 MIR6859-1 LOC100506504 DVL1 LOC105378948 MIR1302-2 LOC105378947 UBE2J2 SSU72 MTND2P28 TTLL10 LOC729737 MIR200A CDK11B KLHL17 PRKCZ LOC102725121 MIR6726
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss MIR6727 ACAP3 DVL1 TAS1R3 PUSL1 CPTP CPSF3L MIR6808 MIR6726
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss MIR6727 AURKAIP1 ACAP3 DVL1 TAS1R3 PUSL1 SCNN1D CPTP CPSF3L MXRA8 MIR6808 MIR6726
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain MIR6727 AURKAIP1 ACAP3 DVL1 TAS1R3 PUSL1 CPTP CPSF3L MXRA8 MIR6808
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss FAM132A AURKAIP1 TAS1R3 TTLL10-AS1 CPTP RNF223 ACAP3 MIR429 TNFRSF18 PUSL1 TNFRSF4 SDF4 MIR6727 DVL1 MXRA8 UBE2J2 TTLL10 B3GALT6 C1orf159 MIR200B LINC01342 SCNN1D MIR200A CPSF3L MIR6808 MIR6726
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss MIR6727 ACAP3 DVL1 TAS1R3 PUSL1 CPTP CPSF3L MXRA8 MIR6808
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion MIR6727 AURKAIP1 ACAP3 DVL1 TAS1R3 PUSL1 SCNN1D CPTP CPSF3L MXRA8 MIR6808 MIR6726
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication FAM132A AGRN TAS1R3 TTLL10-AS1 LOC100288175 CPTP RNF223 ACAP3 MIR429 TNFRSF18 PUSL1 TNFRSF4 SDF4 MIR6727 LOC105378948 UBE2J2 TTLL10 B3GALT6 C1orf159 MIR200B LINC01342 SCNN1D MIR200A CPSF3L MIR6726
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss FAM132A LOC100288069 PLEKHN1 TTLL10-AS1 CPTP ATAD3A MRPL20 RNF223 LINC00115 ACAP3 MIR429 TNFRSF18 PUSL1 TNFRSF4 ATAD3B ANKRD65 FAM41C LOC100287934 MIR6727 PERM1 SLC35E2B ATAD3C LOC107985728 NOC2L LOC107985729 MIB2 MXRA8 CDK11A LOC284600 MMP23B TMEM88B B3GALT6 C1orf159 MIR200B LOC148413 CPSF3L LINC01128 ISG15 MIR6808 FNDC10 AURKAIP1 AGRN TAS1R3 SAMD11 LOC100288175 NADK MMP23A VWA1 CCNL2 FAM87B SDF4 TMEM240 LOC102724312 HES4 DVL1 LOC105378948 UBE2J2 SSU72 TTLL10 SLC35E2 LINC01342 SCNN1D MIR200A CDK11B KLHL17 MIR6726 LOC100130417
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss MIR6727 ACAP3 DVL1 TAS1R3 PUSL1 CPTP CPSF3L MXRA8 MIR6808
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain MIR6727 ACAP3 DVL1 TAS1R3 PUSL1 SCNN1D CPTP CPSF3L MXRA8 MIR6808 MIR6726
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion FAM132A AURKAIP1 TAS1R3 TTLL10-AS1 CPTP ATAD3A MRPL20 ACAP3 VWA1 MIR429 CCNL2 TNFRSF18 PUSL1 TNFRSF4 ATAD3B SDF4 ANKRD65 TMEM240 LOC102724312 MIR6727 DVL1 ATAD3C LOC107985729 MIB2 MXRA8 UBE2J2 SSU72 MMP23B TTLL10 TMEM88B B3GALT6 C1orf159 MIR200B LINC01342 SCNN1D MIR200A CDK11B LOC148413 CPSF3L MIR6808 FNDC10 MIR6726
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss MIR6727 AURKAIP1 DVL1 TAS1R3 CPTP MXRA8 UBE2J2 ACAP3 PUSL1 SCNN1D CPSF3L MIR6808 MIR6726
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain FAM132A AURKAIP1 AGRN TAS1R3 PLEKHN1 TTLL10-AS1 SAMD11 LOC100288175 CPTP MRPL20 RNF223 ACAP3 MIR429 CCNL2 TNFRSF18 PUSL1 TNFRSF4 SDF4 ANKRD65 HES4 MIR6727 PERM1 DVL1 LOC107985728 LOC105378948 NOC2L MXRA8 UBE2J2 LOC284600 TTLL10 TMEM88B B3GALT6 C1orf159 MIR200B LINC01342 SCNN1D MIR200A LOC148413 KLHL17 CPSF3L ISG15 MIR6808 MIR6726 LOC100130417
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR6727 FAM132A TTLL10-AS1 UBE2J2 TTLL10 ACAP3 MIR429 B3GALT6 TNFRSF18 PUSL1 MIR200B TNFRSF4 SCNN1D SDF4 MIR200A CPSF3L MIR6726

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

pseudouridine synthesis
tRNA processing
RNA binding
pseudouridine synthase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0