This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03371589_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147190374 on build GRCh38
Cytoband
1q21.1
Assay ID Hs03371589_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 XP_005273003.1
XM_005272947.4 XP_005273004.1
XM_005272948.4 XP_005273005.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 XP_011507652.1
XM_011509351.2 XP_011507653.1
XM_017000801.1 XP_016856290.1
XM_017000802.1 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
AK314647.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain NOTCH2NL NBPF14 NUDT4P2 MIR6077 PPIAL4G GJA8 RNVU1-8 LOC100132057 BCL9 PDE4DIP NBPF13P LOC105371226 RNU1-13P PDZK1P1 GJA5 LOC728989 RNVU1-6 NBPF12 LOC105371242 LOC101927468 ACP6 CHD1L LINC01138 LINC00624 LOC101928979 RNVU1-3 RNVU1-7 HYDIN2 PDIA3P1 NBPF10 GPR89B FMO5 LOC100996517 LOC100996763 PRKAB2 CH17-408M7.1 RNVU1-1 NUDT4P1 LOC100505824 MIR5087 NBPF11 LOC284561
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss NOTCH2NL NBPF14 NUDT4P2 PPIAL4G GJA8 RNVU1-8 LOC100132057 MIR6736 PDE4DIP NBPF13P RNU1-13P GJA5 HFE2 RNVU1-6 NBPF12 LOC101927468 POLR3GL PEX11B RNVU1-7 HYDIN2 GPR89A POLR3C TXNIP LOC284561 MIR6077 RNF115 BCL9 PIAS3 LOC105371226 PDZK1P1 LOC728989 CD160 LIX1L ANKRD34A LOC105371242 ACP6 CHD1L LINC01138 LINC00624 LOC101928979 RNVU1-3 RBM8A ITGA10 PDZK1 PDIA3P1 ANKRD35 NBPF10 GNRHR2 GPR89B FMO5 LOC100996517 LOC100996763 PRKAB2 CH17-408M7.1 RNVU1-1 NUDT4P1 LOC100505824 NUDT17 MIR5087 NBPF11
dgv67e199 Chr.1:147188956 - 147198583 on Build GRCh38 Deletion CHD1L FMO5
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion LOC728989 CHD1L RNVU1-8 FMO5 PRKAB2 NBPF13P PDIA3P1
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss NOTCH2NL NBPF14 NUDT4P2 MIR6077 PPIAL4G GJA8 RNVU1-8 LOC100132057 BCL9 PDE4DIP NBPF13P LOC105371226 RNU1-13P PDZK1P1 GJA5 LOC728989 NBPF12 LOC105371242 LOC101927468 ACP6 CHD1L LINC01138 LINC00624 RNVU1-3 RNVU1-7 HYDIN2 PDIA3P1 NBPF10 GPR89B FMO5 LOC100996517 LOC100996763 PRKAB2 CH17-408M7.1 RNVU1-1 NUDT4P1 LOC100505824 MIR5087 NBPF11 LOC284561
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion NOTCH2NL NBPF14 NUDT4P2 MIR6077 PPIAL4G GJA8 RNVU1-8 LOC100132057 BCL9 PDE4DIP NBPF13P LOC105371226 RNU1-13P PDZK1P1 GJA5 LOC728989 LIX1L HFE2 ANKRD34A RNVU1-6 NBPF12 LOC105371242 LOC101927468 POLR3GL ACP6 CHD1L LINC01138 LINC00624 LOC101928979 RNVU1-3 RNVU1-7 HYDIN2 PDIA3P1 NBPF10 GPR89B FMO5 TXNIP LOC100996517 LOC100996763 PRKAB2 CH17-408M7.1 RNVU1-1 NUDT4P1 LOC100505824 MIR5087 NBPF11 LOC284561
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication NOTCH2NL NBPF14 NUDT4P2 MIR6077 PPIAL4G GJA8 RNVU1-8 LOC100132057 BCL9 PDE4DIP NBPF13P LOC105371226 RNU1-13P PDZK1P1 GJA5 LOC728989 NBPF12 LOC105371242 LOC101927468 ACP6 CHD1L LINC01138 LINC00624 RNVU1-3 RNVU1-7 HYDIN2 PDIA3P1 GPR89B FMO5 LOC100996517 LOC100996763 PRKAB2 CH17-408M7.1 RNVU1-1 NUDT4P1 LOC100505824 MIR5087 NBPF11 LOC284561
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain NOTCH2NL NBPF14 NUDT4P2 MIR6077 PPIAL4G GJA8 RNVU1-8 LOC100132057 BCL9 PDE4DIP NBPF13P LOC105371226 RNU1-13P PDZK1P1 GJA5 LOC728989 RNVU1-6 NBPF12 LOC105371242 LOC101927468 ACP6 CHD1L LINC01138 LINC00624 LOC101928979 RNVU1-3 RNVU1-7 HYDIN2 PDIA3P1 NBPF10 GPR89B FMO5 LOC100996517 LOC100996763 PRKAB2 CH17-408M7.1 RNVU1-1 NUDT4P1 LOC100505824 MIR5087 NBPF11 LOC284561
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion NOTCH2NL NBPF14 NUDT4P2 MIR6077 PPIAL4G GJA8 RNVU1-8 LOC100132057 BCL9 PDE4DIP NBPF13P LOC105371226 RNU1-13P PDZK1P1 GJA5 LOC728989 NBPF12 LOC105371242 NBPF9 LOC101927468 ACP6 CHD1L LINC01138 LINC00624 LOC101928979 RNVU1-3 RNVU1-7 HYDIN2 PDIA3P1 NBPF10 GPR89B FMO5 LOC100996517 LOC100996763 PRKAB2 CH17-408M7.1 RNVU1-1 NUDT4P1 LOC100505824 MIR5087 NBPF11 LOC284561
esv3587435 Chr.1:147189056 - 147198713 on Build GRCh38 Loss CHD1L FMO5
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss CHD1L FMO5 PRKAB2 PDIA3P1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5jw8n:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0