This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03371881_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1266075 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03371881_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Overlaps Intron 3 - Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss MIR6726 ACAP3 UBE2J2 SCNN1D
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication SDF4 C1orf159 MIR200A MIR200B UBE2J2 FAM132A CPSF3L PUSL1 TNFRSF18 CPTP AGRN MIR6727 MIR6726 LOC105378948 LINC01342 TNFRSF4 TTLL10-AS1 MIR429 RNF223 LOC100288175 ACAP3 B3GALT6 TAS1R3 TTLL10 SCNN1D
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss C1orf159 MMP23A SLC35E2B ATAD3A MIR200A MIR200B CDK11B MIB2 DVL1 MIR6808 HES4 CPTP MIR6726 LOC105378948 ANKRD65 LINC01342 LINC01128 LOC100130417 LOC107985728 PERM1 SSU72 TTLL10-AS1 SLC35E2 TMEM88B CDK11A ISG15 RNF223 VWA1 SAMD11 LOC107985729 TTLL10 LOC284600 ATAD3C SDF4 FAM87B LOC100287934 UBE2J2 FAM132A CPSF3L PUSL1 TNFRSF18 MRPL20 AGRN MIR6727 CCNL2 ATAD3B LOC148413 MMP23B TNFRSF4 NOC2L PLEKHN1 MXRA8 LOC102724312 TMEM240 LOC100288069 MIR429 NADK FNDC10 AURKAIP1 KLHL17 LOC100288175 ACAP3 B3GALT6 TAS1R3 LINC00115 FAM41C SCNN1D
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss C1orf159 ATAD3A MIR200A MIR200B LOC105378592 MIB2 MIR6859-2 LOC105378948 ANKRD65 LOC105378589 LINC01128 OR4F5 PERM1 TTLL10-AS1 SLC35E2 LOC105378947 TMEM88B CDK11A ISG15 RNF223 VWA1 SAMD11 LOC107985729 MTND1P23 ATAD3C LOC105378591 FAM87B UBE2J2 SKI CPSF3L PUSL1 TNFRSF18 ATAD3B GNB1 CFAP74 LOC100506504 NOC2L PLEKHN1 MXRA8 TMEM52 TMEM240 LOC100288069 MIR429 FNDC10 KLHL17 LOC100134822 B3GALT6 WASH7P LINC00115 MMP23A SLC35E2B MIR6723 PRKCZ CDK11B DVL1 MIR6808 HES4 CPTP MIR6726 LOC100129534 LINC01342 LOC100130417 LOC107985728 GABRD SSU72 DDX11L1 MTND2P28 LOC102725121 OR4F16 TTLL10 LOC284600 SDF4 LOC100287934 MORN1 FAM132A MRPL20 AGRN MIR6727 CCNL2 LOC148413 MMP23B TNFRSF4 FAAP20 MIR1302-2 LOC100133331 LOC102724312 OR4F29 FAM138A NADK CALML6 AURKAIP1 LOC100288175 LOC729737 ACAP3 LOC100132287 TAS1R3 LOC105378949 LOC101928626 FAM41C MIR6859-1 LOC107984841 SCNN1D
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss MIR6726 SDF4 ACAP3 B3GALT6 UBE2J2 FAM132A SCNN1D
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss SDF4 C1orf159 FAM87B MIR200A MIR200B UBE2J2 FAM132A TNFRSF18 HES4 AGRN LOC105378948 LINC01342 LINC01128 LOC100130417 TNFRSF4 LOC107985728 NOC2L PERM1 PLEKHN1 TTLL10-AS1 MIR429 ISG15 RNF223 KLHL17 LOC100288175 B3GALT6 SAMD11 LINC00115 TTLL10 FAM41C LOC284600
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion SDF4 C1orf159 ATAD3A MIR200A MIR200B UBE2J2 FAM132A CDK11B CPSF3L MIB2 DVL1 PUSL1 MIR6808 TNFRSF18 MRPL20 CPTP MIR6727 CCNL2 MIR6726 ATAD3B LOC148413 ANKRD65 MMP23B LINC01342 TNFRSF4 MXRA8 LOC102724312 SSU72 TTLL10-AS1 TMEM240 MIR429 TMEM88B FNDC10 AURKAIP1 VWA1 ACAP3 B3GALT6 LOC107985729 TAS1R3 TTLL10 SCNN1D ATAD3C
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss UBE2J2 CPSF3L DVL1 PUSL1 MIR6808 CPTP MIR6727 MIR6726 AURKAIP1 ACAP3 TAS1R3 MXRA8 SCNN1D
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain SDF4 C1orf159 MIR200A MIR200B UBE2J2 FAM132A CPSF3L DVL1 PUSL1 MIR6808 TNFRSF18 MRPL20 HES4 CPTP AGRN MIR6727 CCNL2 MIR6726 LOC148413 LOC105378948 ANKRD65 LINC01342 LOC100130417 TNFRSF4 LOC107985728 NOC2L PERM1 PLEKHN1 MXRA8 TTLL10-AS1 MIR429 TMEM88B ISG15 RNF223 AURKAIP1 KLHL17 LOC100288175 ACAP3 B3GALT6 SAMD11 TAS1R3 TTLL10 SCNN1D LOC284600
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain SDF4 C1orf159 TTLL10-AS1 MIR200A MIR429 MIR200B UBE2J2 FAM132A RNF223 TNFRSF18 LOC105378948 LINC01342 B3GALT6 TNFRSF4 TTLL10 SCNN1D
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss SDF4 C1orf159 TTLL10-AS1 MIR200A MIR429 MIR200B UBE2J2 FAM132A RNF223 TNFRSF18 MIR6726 ACAP3 LINC01342 B3GALT6 TNFRSF4 TTLL10 SCNN1D
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain SDF4 TTLL10-AS1 MIR200A MIR429 MIR200B UBE2J2 FAM132A CPSF3L PUSL1 TNFRSF18 MIR6727 MIR6726 ACAP3 B3GALT6 TNFRSF4 TTLL10 SCNN1D

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-2x6nv:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline