This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03374200_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1625918 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03374200_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Overlaps Intron 8 - Exon 9
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 NP_001164157.1
NM_001170687.1 NP_001164158.1
NM_001170688.1 NP_001164159.1
NM_001170689.1 NP_001164160.1
NM_080875.2 NP_543151.2
NR_033183.1
XM_006710372.1 XP_006710435.1
XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1
XM_017000358.1 XP_016855847.1
XM_017000359.1 XP_016855848.1
XM_017000360.1 XP_016855849.1
XM_017000361.1 XP_016855850.1
XM_017000362.1 XP_016855851.1
XM_017000363.1 XP_016855852.1
XM_017000364.1 XP_016855853.1
XM_017000365.1 XP_016855854.1
XM_017000366.1 XP_016855855.1
AB074480.1 BAB92950.1
AB076691.1 BAC00992.1
AB076692.1 BAC00993.1
AB076693.1 BAC00994.1
AB097000.1 BAC77353.1
AK091610.1 BAC03707.1
AK095914.1 BAC04646.1
AK096295.1 BAC04752.1
AK097106.1 BAC04952.1
AK098785.1 BAC05413.1
AK122751.1
AK122863.1
AK128167.1
AK128553.1 8 1545
AK293642.1
AK302757.1
AK309362.1
AL834527.1 CAD39183.1
BC016490.1 AAH16490.1
BC037542.1 AAH37542.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MIB2 MMP23A SLC35E2 NADK CDK11A GNB1 CDK11B FNDC10 SLC35E2B MMP23B
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss MIB2 MMP23A SLC35E2 CDK11A CDK11B FNDC10 SLC35E2B MMP23B SSU72
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain MIB2 MMP23A CDK11A CDK11B SLC35E2B MMP23B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss NOC2L MRPL20 RNF223 CPTP SCNN1D MIR200A SDF4 CDK11A C1orf159 FAM87B LINC01128 PERM1 MMP23B MXRA8 DVL1 B3GALT6 NADK AGRN TNFRSF18 LOC107985728 SSU72 LOC284600 LOC100287934 MIR429 SAMD11 SLC35E2B LOC102724312 FAM132A VWA1 FAM41C HES4 LINC00115 PUSL1 ISG15 CDK11B LOC100130417 TTLL10-AS1 MIR6808 LOC100288069 ATAD3A CCNL2 LOC105378948 PLEKHN1 MIR6727 LOC107985729 UBE2J2 MIR200B CPSF3L TMEM240 MIB2 KLHL17 SLC35E2 LINC01342 TAS1R3 ANKRD65 LOC148413 TMEM88B MMP23A MIR6726 ATAD3C TNFRSF4 LOC100288175 TTLL10 ATAD3B FNDC10 AURKAIP1 ACAP3
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss NOC2L MRPL20 TMEM52 OR4F16 LOC729737 RNF223 PRKCZ MORN1 LOC105378947 MIR6859-2 CFAP74 LINC01128 PERM1 DVL1 B3GALT6 OR4F5 AGRN LOC107984841 SSU72 LOC100287934 MIR429 MIR1302-2 CALML6 ISG15 CDK11B FAAP20 LOC100134822 DDX11L1 ATAD3A LOC100506504 OR4F29 CCNL2 LOC105378948 GNB1 MIR6727 LOC107985729 UBE2J2 TMEM240 GABRD MIB2 KLHL17 TMEM88B MIR6723 MIR6726 TNFRSF4 AURKAIP1 ACAP3 LOC100129534 CPTP MTND1P23 SCNN1D MIR6859-1 MIR200A SDF4 CDK11A C1orf159 FAM87B LOC100132287 MMP23B MXRA8 WASH7P NADK TNFRSF18 LOC107985728 LOC105378949 LOC284600 LOC100133331 SAMD11 SLC35E2B LOC102724312 FAM132A VWA1 FAM41C HES4 LINC00115 PUSL1 LOC105378589 LOC105378591 LOC100130417 TTLL10-AS1 MIR6808 LOC100288069 MTND2P28 PLEKHN1 MIR200B CPSF3L LOC101928626 SKI SLC35E2 LINC01342 TAS1R3 ANKRD65 LOC148413 LOC105378592 LOC102725121 MMP23A ATAD3C FAM138A LOC100288175 TTLL10 ATAD3B FNDC10
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MIB2 MMP23A SLC35E2 NADK CDK11A GNB1 CDK11B FNDC10 SLC35E2B MMP23B SSU72
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MRPL20 PUSL1 CPTP CDK11B TTLL10-AS1 SCNN1D MIR6808 ATAD3A MIR200A CCNL2 SDF4 MIR6727 C1orf159 LOC107985729 UBE2J2 MMP23B MXRA8 MIR200B DVL1 CPSF3L TMEM240 MIB2 B3GALT6 LINC01342 TAS1R3 ANKRD65 LOC148413 TNFRSF18 SSU72 TMEM88B MIR6726 ATAD3C TNFRSF4 MIR429 TTLL10 ATAD3B FNDC10 LOC102724312 FAM132A AURKAIP1 VWA1 ACAP3
esv2661270 Chr.1:1625365 - 1636030 on Build GRCh38 Deletion MIB2 CDK11B MMP23B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss MIB2 MMP23A SLC35E2 NADK CDK11A CDK11B FNDC10 SLC35E2B MMP23B SSU72
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss MIB2 CDK11B SLC35E2B MMP23B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss MRPL20 TMEM52 PRKCZ LOC105378589 LOC105378591 CDK11B CFAP74 ATAD3A CCNL2 CDK11A GNB1 LOC107985729 MMP23B TMEM240 GABRD MIB2 SLC35E2 NADK ANKRD65 LOC148413 SSU72 TMEM88B MMP23A LOC105378949 ATAD3C ATAD3B FNDC10 SLC35E2B LOC102724312 VWA1 CALML6

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0