This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03374407_cn
See other SLC35E2B CNV Assays ›
Gene Symbol
SLC35E2B
Assay Reference Genome
Location

Chr.1:1681119 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03374407_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728661

Gene Name:

solute carrier family 35 member E2B

Gene Aliases:

SLC35E2

Location:

Chr.1:1661478-1692804 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC35E2B NM_001110781.2 NP_001104251.1
NM_001290264.1 NP_001277193.1
XM_011542072.1 XP_011540374.1
AB007916.3
AK125619.1
AK307886.1
BC110653.1 AAI10654.1
BG328537.1
CR936618.1 CAI56761.1
DA794179.1
DB450858.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n54 Chr.1:1663759 - 1696548 on Build GRCh38 Loss SLC35E2B
esv3585012 Chr.1:1677771 - 1684388 on Build GRCh38 Gain SLC35E2B
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion FNDC10 GNB1 CDK11A CDK11B SLC35E2B SLC35E2 NADK MIB2 MMP23B MMP23A
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss FNDC10 CDK11A CDK11B SLC35E2B SSU72 SLC35E2 MIB2 MMP23B MMP23A
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain CDK11A CDK11B SLC35E2B MIB2 MMP23B MMP23A
esv3585010 Chr.1:1670280 - 1751138 on Build GRCh38 Loss CDK11A SLC35E2B SLC35E2 MMP23A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss ATAD3C PRKCZ ISG15 UBE2J2 TMEM52 SSU72 KLHL17 FAM138A NOC2L LOC105378949 PUSL1 MRPL20 MTND1P23 TTLL10-AS1 MIR6723 MIR6726 ANKRD65 VWA1 LOC284600 GABRD MIR200A LOC105378591 MIR1302-2 B3GALT6 ATAD3A OR4F5 MIR6859-2 FAM87B LINC00115 LOC100130417 MMP23B LOC729737 LOC101928626 MIR6859-1 MORN1 LOC105378592 LOC100129534 MIR6727 AURKAIP1 DVL1 MIR429 MXRA8 LOC107985729 TNFRSF4 LOC100133331 LOC105378589 SAMD11 NADK FNDC10 PLEKHN1 LOC100132287 LOC102725121 TMEM240 OR4F16 LINC01128 LOC100288175 MIR200B DDX11L1 RNF223 SDF4 C1orf159 CDK11B MIR6808 MIB2 FAM132A ACAP3 LOC100506504 SCNN1D LOC102724312 SLC35E2 LOC100288069 PERM1 MMP23A SKI TNFRSF18 GNB1 CFAP74 LOC100287934 LOC148413 SLC35E2B CCNL2 WASH7P LINC01342 TAS1R3 LOC107985728 TTLL10 FAM41C CDK11A OR4F29 MTND2P28 CPTP ATAD3B TMEM88B LOC105378947 CALML6 LOC100134822 LOC105378948 FAAP20 AGRN LOC107984841 CPSF3L HES4
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication FNDC10 GNB1 CDK11A CDK11B SLC35E2B SSU72 SLC35E2 NADK MIB2 MMP23B MMP23A
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11A CDK11B SLC35E2B MMP23A
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss FNDC10 CDK11A CDK11B SLC35E2B SSU72 SLC35E2 NADK MIB2 MMP23B MMP23A
esv3585007 Chr.1:1647187 - 1714507 on Build GRCh38 Loss CDK11A CDK11B SLC35E2B MMP23A
nsv950516 Chr.1:1663141 - 1715961 on Build GRCh38 Deletion CDK11A SLC35E2B MMP23A
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain CDK11A CDK11B SLC35E2B SLC35E2 NADK MMP23B MMP23A
nsv545022 Chr.1:1665581 - 1683909 on Build GRCh38 Loss SLC35E2B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss C1orf159 ATAD3C ISG15 UBE2J2 CDK11B SSU72 MIR6808 KLHL17 MIB2 NOC2L FAM132A PUSL1 ACAP3 SCNN1D MRPL20 LOC102724312 TTLL10-AS1 SLC35E2 LOC100288069 PERM1 MMP23A MIR6726 TNFRSF18 ANKRD65 VWA1 LOC100287934 LOC284600 MIR200A LOC148413 SLC35E2B CCNL2 B3GALT6 LINC01342 TAS1R3 ATAD3A FAM87B LOC107985728 LINC00115 TTLL10 FAM41C LOC100130417 MMP23B MIR6727 AURKAIP1 DVL1 MIR429 MXRA8 LOC107985729 TNFRSF4 CDK11A SAMD11 NADK FNDC10 CPTP ATAD3B PLEKHN1 TMEM88B TMEM240 LOC105378948 AGRN LINC01128 LOC100288175 CPSF3L HES4 MIR200B RNF223 SDF4
nsv545026 Chr.1:1671348 - 1706566 on Build GRCh38 Loss CDK11A SLC35E2B MMP23A
nsv545018 Chr.1:1638499 - 1688102 on Build GRCh38 Loss CDK11B SLC35E2B
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain CDK11A CDK11B SLC35E2B SLC35E2 MMP23A
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss ATAD3C PRKCZ CDK11B TMEM52 SSU72 MIB2 LOC105378949 LOC107985729 MRPL20 CDK11A LOC102724312 LOC105378589 SLC35E2 NADK MMP23A FNDC10 ANKRD65 GNB1 ATAD3B CFAP74 TMEM88B VWA1 CALML6 GABRD TMEM240 LOC148413 SLC35E2B LOC105378591 CCNL2 ATAD3A MMP23B

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0