This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03376086_cn
See other SLC35E2B CNV Assays ›
Gene Symbol
SLC35E2B
Assay Reference Genome
Location

Chr.1:1669834 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03376086_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728661

Gene Name:

solute carrier family 35 member E2B

Gene Aliases:

SLC35E2

Location:

Chr.1:1661478-1692804 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC35E2B NM_001110781.2 NP_001104251.1
NM_001290264.1 NP_001277193.1
XM_011542072.1 XP_011540374.1
AB007916.3
AK092311.1 1 306
AK125619.1
AK307886.1
BC110653.1 AAI10654.1
CR936618.1 CAI56761.1
DA794179.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n54 Chr.1:1663759 - 1696548 on Build GRCh38 Loss SLC35E2B
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MMP23B CDK11A FNDC10 SLC35E2B MIB2 CDK11B MMP23A GNB1 SLC35E2 NADK
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss SSU72 MMP23B CDK11A FNDC10 SLC35E2B MIB2 CDK11B MMP23A SLC35E2
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain MMP23B CDK11A SLC35E2B MIB2 CDK11B MMP23A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC107985729 RNF223 LOC100288069 NOC2L FAM132A KLHL17 CDK11A LOC100133331 TMEM52 TNFRSF18 LOC105378949 MMP23A MIR200B LOC284600 LOC148413 LOC100132287 B3GALT6 SCNN1D OR4F5 ATAD3A TMEM240 ACAP3 MTND2P28 TTLL10 PRKCZ PLEKHN1 MORN1 FAM138A GABRD VWA1 SKI SSU72 LOC102725121 OR4F29 LOC729737 FAM41C NADK MIR6859-2 MIR200A DDX11L1 MIR6808 SDF4 LOC105378592 GNB1 SLC35E2 LINC01342 LOC105378948 LOC107985728 LOC105378591 MIR6726 MTND1P23 CFAP74 LOC100134822 LOC100130417 MIB2 AURKAIP1 MIR429 MIR6859-1 CCNL2 LOC100506504 LOC101928626 MRPL20 TAS1R3 C1orf159 AGRN DVL1 TTLL10-AS1 ATAD3C LOC100129534 MMP23B LINC00115 TNFRSF4 HES4 PUSL1 LOC105378589 LOC105378947 UBE2J2 LOC102724312 MIR1302-2 LOC100287934 MXRA8 SAMD11 LOC107984841 FNDC10 ISG15 LOC100288175 CALML6 WASH7P CPSF3L CDK11B FAAP20 OR4F16 MIR6723 SLC35E2B TMEM88B ATAD3B MIR6727 PERM1 ANKRD65 FAM87B LINC01128 CPTP
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SSU72 MMP23B CDK11A FNDC10 SLC35E2B MIB2 CDK11B MMP23A GNB1 SLC35E2 NADK
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11A SLC35E2B CDK11B MMP23A
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss SSU72 MMP23B CDK11A FNDC10 SLC35E2B MIB2 CDK11B MMP23A SLC35E2 NADK
esv3585008 Chr.1:1663929 - 1681002 on Build GRCh38 Loss SLC35E2B
esv3585007 Chr.1:1647187 - 1714507 on Build GRCh38 Loss CDK11A SLC35E2B CDK11B MMP23A
nsv950516 Chr.1:1663141 - 1715961 on Build GRCh38 Deletion CDK11A SLC35E2B MMP23A
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain MMP23B CDK11A SLC35E2B CDK11B MMP23A SLC35E2 NADK
nsv545022 Chr.1:1665581 - 1683909 on Build GRCh38 Loss SLC35E2B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC107985729 RNF223 LOC100130417 LOC100288069 MIB2 NOC2L AURKAIP1 FAM132A MIR429 CCNL2 MRPL20 TAS1R3 KLHL17 C1orf159 CDK11A AGRN TNFRSF18 DVL1 TTLL10-AS1 MMP23A MIR200B ATAD3C LOC284600 LOC148413 MMP23B LINC00115 TNFRSF4 B3GALT6 SCNN1D HES4 ATAD3A PUSL1 TMEM240 ACAP3 UBE2J2 LOC102724312 TTLL10 LOC100287934 MXRA8 PLEKHN1 SAMD11 FNDC10 ISG15 LOC100288175 VWA1 SSU72 CPSF3L CDK11B FAM41C NADK MIR200A MIR6808 SDF4 SLC35E2B TMEM88B ATAD3B MIR6727 PERM1 ANKRD65 SLC35E2 LINC01342 FAM87B LOC105378948 LOC107985728 LINC01128 CPTP MIR6726
nsv545018 Chr.1:1638499 - 1688102 on Build GRCh38 Loss SLC35E2B CDK11B
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain CDK11A SLC35E2B CDK11B MMP23A SLC35E2
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss PRKCZ CFAP74 LOC107985729 FNDC10 MIB2 GABRD CALML6 VWA1 CCNL2 MRPL20 SSU72 CDK11A TMEM52 LOC105378949 CDK11B MMP23A ATAD3C NADK LOC148413 MMP23B SLC35E2B TMEM88B ATAD3B ATAD3A ANKRD65 LOC105378589 GNB1 SLC35E2 TMEM240 LOC102724312 LOC105378591

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline