This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03381543_cn
See other SDF4 CNV Assays ›
Gene Symbol
SDF4
Assay Reference Genome
Location

Chr.1:1221168 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03381543_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51150

Gene Name:

stromal cell derived factor 4

Gene Aliases:

Cab45, SDF-4

Location:

Chr.1:1216908-1232067 on Build GRCh38

Assay Gene Location:

Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SDF4 NM_016176.3 NP_057260.2
NM_016547.2 NP_057631.1
XM_011541556.1 XP_011539858.1
AF132749.1 AAL75950.1
AF153686.1 AAD51612.1
AF178986.1 AAF44350.1
AK000717.1
AK027277.1 BAB55012.1
AK075352.1 BAC11563.1
AK126927.1 1 184
AK222521.1 BAD96241.1
AK223079.1 BAD96799.1
AK299810.1
AK315517.1
BC006211.2 AAH06211.1
BC007625.1 AAH07625.1
BC008917.2 AAH08917.1
BC011244.1 AAH11244.1
BC022375.1 AAH22375.1
L79912.1 AAL40084.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv159887 Chr.1:1217713 - 1222580 on Build GRCh38 Deletion SDF4
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication CPTP TNFRSF4 TTLL10 PUSL1 TAS1R3 SDF4 TNFRSF18 FAM132A MIR6727 RNF223 AGRN LINC01342 MIR200B LOC100288175 C1orf159 SCNN1D ACAP3 TTLL10-AS1 B3GALT6 UBE2J2 MIR429 CPSF3L LOC105378948 MIR200A MIR6726
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CPTP MIB2 MMP23A ATAD3C NOC2L CDK11B PLEKHN1 DVL1 TTLL10 PUSL1 TAS1R3 SSU72 SDF4 TNFRSF18 VWA1 AURKAIP1 MIR6727 ATAD3B PERM1 RNF223 MIR6808 LOC102724312 LINC01342 MIR200B FAM87B SCNN1D LOC100288069 ACAP3 FAM41C B3GALT6 UBE2J2 CCNL2 LINC01128 LOC100287934 SLC35E2B MIR200A MIR6726 MRPL20 MMP23B FNDC10 HES4 TMEM88B TNFRSF4 LINC00115 LOC107985729 FAM132A SLC35E2 ANKRD65 AGRN TMEM240 LOC100288175 C1orf159 NADK LOC148413 LOC284600 LOC100130417 TTLL10-AS1 KLHL17 ISG15 LOC107985728 MIR429 CPSF3L MXRA8 SAMD11 LOC105378948 CDK11A ATAD3A
nsv455661 Chr.1:1217251 - 1275912 on Build GRCh38 Loss B3GALT6 UBE2J2 SDF4 FAM132A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss OR4F16 MMP23A LOC100132287 FAM138A ATAD3C PLEKHN1 TNFRSF18 AURKAIP1 MIR6727 ATAD3B OR4F29 MTND1P23 PERM1 RNF223 MIR6808 LOC102724312 MIR6859-2 LINC01342 LOC105378947 LOC105378589 LOC100288069 GNB1 UBE2J2 LOC100287934 OR4F5 LOC729737 MIR6726 MMP23B FNDC10 LOC105378592 TMEM88B PRKCZ LINC00115 FAM132A LOC100134822 TMEM52 AGRN TMEM240 MIR6859-1 LOC100288175 C1orf159 NADK LOC101928626 LOC100130417 TTLL10-AS1 LOC100129534 ISG15 CPSF3L MXRA8 CALML6 LOC105378948 WASH7P LOC107984841 CPTP MIB2 NOC2L CDK11B DVL1 TTLL10 PUSL1 TAS1R3 SSU72 LOC100506504 SDF4 LOC100133331 VWA1 MIR6723 LOC102725121 MORN1 MIR200B FAM87B SCNN1D MIR1302-2 LOC105378949 ACAP3 MTND2P28 FAM41C B3GALT6 CCNL2 LINC01128 SLC35E2B SKI LOC105378591 MIR200A MRPL20 HES4 TNFRSF4 LOC107985729 CFAP74 SLC35E2 ANKRD65 GABRD FAAP20 LOC148413 LOC284600 DDX11L1 KLHL17 LOC107985728 MIR429 SAMD11 CDK11A ATAD3A
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MMP23B FNDC10 CPTP MIB2 TMEM88B ATAD3C TNFRSF4 CDK11B DVL1 TTLL10 PUSL1 TAS1R3 SSU72 SDF4 TNFRSF18 VWA1 AURKAIP1 LOC107985729 FAM132A MIR6727 ATAD3B MIR6808 LOC102724312 ANKRD65 TMEM240 LINC01342 MIR200B C1orf159 SCNN1D LOC148413 ACAP3 TTLL10-AS1 B3GALT6 UBE2J2 CCNL2 MIR429 CPSF3L MXRA8 ATAD3A MIR200A MIR6726 MRPL20
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss HES4 NOC2L TNFRSF4 PLEKHN1 TTLL10 LINC00115 SDF4 TNFRSF18 FAM132A PERM1 RNF223 AGRN LINC01342 MIR200B LOC100288175 FAM87B C1orf159 LOC284600 LOC100130417 TTLL10-AS1 FAM41C KLHL17 ISG15 B3GALT6 UBE2J2 LOC107985728 LINC01128 MIR429 SAMD11 LOC105378948 MIR200A
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain CPTP HES4 TMEM88B NOC2L TNFRSF4 PLEKHN1 DVL1 TTLL10 PUSL1 TAS1R3 SDF4 TNFRSF18 AURKAIP1 FAM132A MIR6727 PERM1 RNF223 MIR6808 ANKRD65 AGRN LINC01342 MIR200B LOC100288175 C1orf159 SCNN1D LOC148413 LOC284600 ACAP3 LOC100130417 TTLL10-AS1 KLHL17 ISG15 B3GALT6 UBE2J2 CCNL2 LOC107985728 MIR429 CPSF3L MXRA8 SAMD11 LOC105378948 MIR200A MIR6726 MRPL20
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TNFRSF4 TTLL10 SDF4 TNFRSF18 FAM132A TTLL10-AS1 B3GALT6 UBE2J2 MIR429 RNF223 LOC105378948 LINC01342 MIR200B MIR200A C1orf159 SCNN1D
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss CPTP TNFRSF4 DVL1 TTLL10 PUSL1 TAS1R3 SDF4 TNFRSF18 AURKAIP1 FAM132A MIR6727 RNF223 MIR6808 LINC01342 MIR200B C1orf159 SCNN1D ACAP3 TTLL10-AS1 B3GALT6 UBE2J2 MIR429 CPSF3L MXRA8 MIR200A MIR6726
nsv544956 Chr.1:1171093 - 1241217 on Build GRCh38 Loss B3GALT6 TNFRSF4 TTLL10 SDF4 TNFRSF18 TTLL10-AS1
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TNFRSF4 TTLL10 PUSL1 SDF4 TNFRSF18 ACAP3 FAM132A MIR6727 TTLL10-AS1 B3GALT6 UBE2J2 MIR429 CPSF3L MIR200B MIR200A MIR6726 SCNN1D

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

UV protection
calcium ion regulated exocytosis
cerebellum development
fat cell differentiation
response to ethanol
zymogen granule exocytosis
calcium ion binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0