This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03382626_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1269042 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03382626_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_017000238.1 XP_016855727.1
AF296658.1 AAK52609.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM794599.1
BM926567.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss MIR6726 SCNN1D ACAP3 UBE2J2
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication B3GALT6 ACAP3 MIR200A TNFRSF4 CPTP AGRN FAM132A TNFRSF18 MIR6727 CPSF3L PUSL1 MIR200B LOC100288175 TAS1R3 TTLL10-AS1 MIR429 LINC01342 SDF4 TTLL10 C1orf159 UBE2J2 MIR6726 RNF223 SCNN1D LOC105378948
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LINC00115 TMEM240 ACAP3 NOC2L TNFRSF4 MIR6808 FAM132A TNFRSF18 PUSL1 VWA1 MIR200B ATAD3A LOC100287934 MRPL20 MMP23B MIB2 LOC148413 NADK MIR429 CCNL2 LOC284600 UBE2J2 ATAD3B ATAD3C SCNN1D LOC100288069 AURKAIP1 FAM41C LOC105378948 HES4 SLC35E2 FNDC10 B3GALT6 SAMD11 MIR200A CPTP AGRN DVL1 LOC102724312 MIR6727 CDK11B ANKRD65 CDK11A CPSF3L TMEM88B KLHL17 SSU72 FAM87B LOC100288175 LOC107985728 TAS1R3 TTLL10-AS1 LOC100130417 MXRA8 LINC01342 MMP23A SDF4 TTLL10 C1orf159 MIR6726 PERM1 RNF223 PLEKHN1 LINC01128 ISG15 LOC107985729 SLC35E2B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss ACAP3 MTND1P23 MIR6808 FAM132A PUSL1 LOC101928626 PRKCZ MIB2 MIR6723 MIR429 CCNL2 LOC102725121 LOC284600 UBE2J2 LOC100288069 WASH7P TMEM52 FAM138A HES4 CPTP LOC100132287 DVL1 MIR6859-1 CDK11B ANKRD65 CDK11A CPSF3L KLHL17 LOC100288175 TAS1R3 OR4F5 LOC105378592 LOC100134822 MXRA8 LINC01342 MMP23A TTLL10 PERM1 PLEKHN1 ISG15 SLC35E2B LINC00115 CALML6 TMEM240 LOC105378947 NOC2L GABRD TNFRSF4 TNFRSF18 LOC100506504 MTND2P28 VWA1 MIR200B SKI ATAD3A LOC100287934 MRPL20 MMP23B LOC100129534 LOC148413 NADK FAAP20 LOC729737 LOC105378949 ATAD3B ATAD3C SCNN1D AURKAIP1 FAM41C LOC105378589 LOC105378948 MIR6859-2 SLC35E2 FNDC10 LOC107984841 OR4F29 MIR1302-2 B3GALT6 OR4F16 SAMD11 MIR200A AGRN LOC102724312 MIR6727 TMEM88B SSU72 FAM87B LOC107985728 DDX11L1 GNB1 TTLL10-AS1 LOC100130417 SDF4 C1orf159 MIR6726 RNF223 LINC01128 MORN1 LOC107985729 LOC100133331 CFAP74 LOC105378591
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss MIR6726 SCNN1D B3GALT6 ACAP3 SDF4 FAM132A UBE2J2
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LINC00115 B3GALT6 NOC2L SAMD11 MIR200A TNFRSF4 AGRN FAM132A TNFRSF18 KLHL17 MIR200B FAM87B LOC100288175 LOC107985728 TTLL10-AS1 MIR429 LOC100130417 LINC01342 LOC284600 SDF4 TTLL10 C1orf159 UBE2J2 PERM1 RNF223 PLEKHN1 LINC01128 FAM41C ISG15 LOC105378948 HES4
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion TMEM240 B3GALT6 ACAP3 MIR200A TNFRSF4 CPTP MIR6808 FAM132A DVL1 LOC102724312 TNFRSF18 MIR6727 CDK11B ANKRD65 CPSF3L PUSL1 TMEM88B SSU72 VWA1 MIR200B ATAD3A MRPL20 MMP23B TAS1R3 MIB2 LOC148413 TTLL10-AS1 MIR429 CCNL2 MXRA8 LINC01342 SDF4 TTLL10 C1orf159 UBE2J2 ATAD3B MIR6726 ATAD3C SCNN1D AURKAIP1 LOC107985729 FNDC10
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss ACAP3 MXRA8 CPTP MIR6808 UBE2J2 DVL1 MIR6726 MIR6727 SCNN1D AURKAIP1 CPSF3L PUSL1 TAS1R3
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain B3GALT6 ACAP3 NOC2L SAMD11 MIR200A TNFRSF4 CPTP AGRN MIR6808 FAM132A DVL1 TNFRSF18 MIR6727 ANKRD65 CPSF3L PUSL1 TMEM88B KLHL17 MIR200B MRPL20 LOC100288175 LOC107985728 TAS1R3 LOC148413 TTLL10-AS1 MIR429 CCNL2 LOC100130417 MXRA8 LINC01342 LOC284600 SDF4 TTLL10 C1orf159 UBE2J2 MIR6726 PERM1 RNF223 PLEKHN1 SCNN1D AURKAIP1 ISG15 LOC105378948 HES4
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TTLL10-AS1 MIR429 B3GALT6 LINC01342 MIR200A TNFRSF4 SDF4 TTLL10 C1orf159 FAM132A UBE2J2 TNFRSF18 RNF223 SCNN1D LOC105378948 MIR200B
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss TTLL10-AS1 MIR429 B3GALT6 ACAP3 LINC01342 MIR200A TNFRSF4 SDF4 TTLL10 C1orf159 FAM132A UBE2J2 MIR6726 TNFRSF18 RNF223 SCNN1D MIR200B
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TTLL10-AS1 MIR429 B3GALT6 ACAP3 MIR200A TNFRSF4 SDF4 TTLL10 FAM132A UBE2J2 MIR6726 TNFRSF18 MIR6727 SCNN1D CPSF3L PUSL1 MIR200B

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline