This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03382801_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:946710 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03382801_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Within Intron 14
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK022756.1 BAB14230.1
AK024284.1
AK092491.1 1 351 BAC03905.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain PERM1 C1orf159 LOC107985728 NOC2L AGRN LOC100288175 ISG15 LOC105378948 LOC100130417 RNF223 PLEKHN1 KLHL17 HES4 LOC284600 SAMD11
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss AURKAIP1 PERM1 MRPL20 TNFRSF18 DVL1 TNFRSF4 MIR200B PRKCZ FAM41C RNF223 ATAD3A KLHL17 MTND1P23 LOC284600 MIR6726 SSU72 PUSL1 MXRA8 TTLL10-AS1 CFAP74 LOC100288069 ACAP3 SDF4 LINC01342 FAM132A CDK11A LOC148413 CCNL2 LOC107985728 GNB1 NADK WASH7P LOC100130417 PLEKHN1 OR4F29 LOC105378591 UBE2J2 LOC102724312 GABRD ANKRD65 LINC00115 MMP23B MTND2P28 SCNN1D TAS1R3 DDX11L1 OR4F16 FAM87B LOC107985729 TTLL10 B3GALT6 MIR6859-1 FAM138A C1orf159 LOC105378947 VWA1 NOC2L AGRN LOC105378948 LOC100134822 LOC100133331 MIR6859-2 LOC105378949 LOC105378589 MMP23A MIR429 HES4 LOC100129534 TMEM52 ATAD3B SAMD11 SLC35E2 LINC01128 LOC102725121 CDK11B LOC105378592 LOC100132287 CPSF3L MIR6727 FAAP20 MIR6808 TMEM240 MIR6723 MIB2 SLC35E2B MIR200A OR4F5 MIR1302-2 LOC101928626 ATAD3C LOC100506504 MORN1 LOC729737 LOC100288175 ISG15 LOC100287934 CPTP LOC107984841 CALML6 FNDC10 SKI TMEM88B
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion PERM1 C1orf159 LOC107985728 NOC2L AGRN LOC100288175 ISG15 LOC105378948 LOC100130417 RNF223 PLEKHN1 KLHL17 HES4 LOC284600 SAMD11
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion PERM1 PLEKHN1 LOC107985728 NOC2L KLHL17 LOC284600 SAMD11 LOC100130417
dgv9n54 Chr.1:925754 - 952840 on Build GRCh38 Loss NOC2L SAMD11
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain PERM1 C1orf159 LOC107985728 NOC2L AGRN TNFRSF18 LOC105378948 TNFRSF4 LOC100130417 MIR200B FAM41C RNF223 PLEKHN1 MIR200A KLHL17 MIR429 HES4 LOC284600 SAMD11 LOC100288175 TTLL10-AS1 ISG15 SDF4 LINC01342 TTLL10
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain PERM1 C1orf159 LOC105378947 LOC107985728 NOC2L AGRN LOC105378948 LOC100130417 FAM41C RNF223 LOC100133331 PLEKHN1 KLHL17 HES4 MTND1P23 LOC101928626 LOC284600 SAMD11 LINC01128 LINC00115 MTND2P28 LOC100288175 ISG15 LOC100287934 LOC100288069 LOC107984841 OR4F16 FAM87B MIR6723
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss AURKAIP1 PERM1 C1orf159 VWA1 MRPL20 NOC2L AGRN TNFRSF18 LOC105378948 DVL1 TNFRSF4 MIR200B FAM41C RNF223 ATAD3A MMP23A KLHL17 MIR429 HES4 ATAD3B LOC284600 SAMD11 MIR6726 SLC35E2 LINC01128 SSU72 CDK11B PUSL1 MXRA8 TTLL10-AS1 LOC100288069 CPSF3L ACAP3 MIR6727 MIR6808 TMEM240 SDF4 LINC01342 FAM132A CDK11A LOC148413 CCNL2 MIB2 LOC107985728 NADK SLC35E2B LOC100130417 PLEKHN1 MIR200A UBE2J2 LOC102724312 ATAD3C ANKRD65 LINC00115 MMP23B LOC100288175 ISG15 LOC100287934 SCNN1D CPTP TAS1R3 FNDC10 TMEM88B FAM87B LOC107985729 TTLL10 B3GALT6
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss PERM1 C1orf159 LOC107985728 NOC2L AGRN TNFRSF18 LOC105378948 TNFRSF4 LOC100130417 MIR200B FAM41C RNF223 PLEKHN1 MIR200A UBE2J2 KLHL17 MIR429 HES4 LOC284600 SAMD11 LINC01128 LINC00115 LOC100288175 TTLL10-AS1 ISG15 FAM87B SDF4 LINC01342 FAM132A TTLL10 B3GALT6
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain PERM1 C1orf159 NOC2L AGRN LOC100288175 ISG15 LOC105378948 RNF223 PLEKHN1 KLHL17 LINC01342 HES4 SAMD11
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain AURKAIP1 PERM1 C1orf159 LOC148413 CCNL2 LOC107985728 MRPL20 NOC2L AGRN TNFRSF18 LOC105378948 DVL1 TNFRSF4 LOC100130417 MIR200B RNF223 PLEKHN1 MIR200A UBE2J2 KLHL17 MIR429 HES4 LOC284600 SAMD11 MIR6726 ANKRD65 PUSL1 MXRA8 LOC100288175 TTLL10-AS1 ISG15 SCNN1D CPTP CPSF3L ACAP3 TAS1R3 MIR6727 TMEM88B MIR6808 SDF4 LINC01342 FAM132A TTLL10 B3GALT6
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss PERM1 PLEKHN1 LOC107985728 NOC2L KLHL17 AGRN HES4 ISG15 LOC284600 SAMD11 LOC100130417
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion PERM1 C1orf159 NOC2L AGRN TNFRSF18 LOC100288175 TTLL10-AS1 ISG15 LOC105378948 TNFRSF4 MIR200B RNF223 PLEKHN1 MIR200A KLHL17 MIR429 SDF4 LINC01342 HES4 TTLL10 SAMD11
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain PERM1 FAM41C PLEKHN1 LOC107985728 NOC2L KLHL17 HES4 LOC284600 SAMD11 LOC100130417 LINC01128

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline