This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03386861_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1798205 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03386861_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion FNDC10 CDK11B CDK11A MMP23B SLC35E2B MMP23A NADK MIB2 GNB1 SLC35E2
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss CDK11A MMP23A NADK GNB1 SLC35E2
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication FNDC10 CDK11B CDK11A SSU72 MMP23B SLC35E2B MMP23A NADK MIB2 GNB1 SLC35E2
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss UBE2J2 MIR200B PLEKHN1 TMEM88B TNFRSF4 OR4F29 LOC105378589 LOC284600 LOC100506504 OR4F5 MTND1P23 LOC107985728 FAM87B LOC100288069 MIR200A LOC102724312 RNF223 C1orf159 DVL1 LOC100130417 ACAP3 TTLL10-AS1 CCNL2 AGRN SDF4 GNB1 LOC107984841 FAAP20 NOC2L SAMD11 TNFRSF18 SLC35E2B TTLL10 MIR6859-2 LINC01128 SKI PRKCZ MIR6726 MIR429 CALML6 HES4 LOC102725121 PERM1 B3GALT6 LOC107985729 DDX11L1 ANKRD65 LOC101928626 LOC100134822 VWA1 CPSF3L SLC35E2 SSU72 MMP23A LOC105378591 GABRD WASH7P LOC100132287 FAM138A LOC100288175 LOC105378949 MMP23B MIR6808 LOC105378592 TAS1R3 TMEM240 CDK11A AURKAIP1 NADK PUSL1 MIR6859-1 ATAD3C FNDC10 LOC148413 MIR1302-2 MXRA8 KLHL17 LOC100129534 ATAD3B LINC00115 LOC100287934 ISG15 CDK11B CFAP74 OR4F16 LOC105378947 MIR6723 LOC105378948 ATAD3A MTND2P28 SCNN1D MIB2 TMEM52 MIR6727 LOC729737 LOC100133331 LINC01342 FAM132A MORN1 CPTP FAM41C MRPL20
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication LOC105378949 NADK GNB1
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss CDK11B SSU72 CFAP74 MMP23A CCNL2 LOC105378591 GABRD ATAD3A LOC105378949 MMP23B TMEM88B MIB2 TMEM52 GNB1 LOC105378589 TMEM240 CDK11A SLC35E2B NADK PRKCZ CALML6 ATAD3C FNDC10 LOC102724312 LOC148413 LOC107985729 ANKRD65 MRPL20 ATAD3B VWA1 SLC35E2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline