Assay Details
Target Gene Details
Entrez Gene ID: | 3777 |
Gene Name: | potassium two pore domain channel subfamily K member 3 |
Gene Aliases: |
K2p3.1, OAT1, PPH4, TASK, TASK-1, TBAK1 |
Location: |
Chr.2:26692713-26733420 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK3 | NM_002246.2 | NP_002237.1 | ||
| AF006823.1 | AAC51777.1 | |||
| AF065163.1 | AAG29340.1 | |||
| BC140701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508833 | Chr.2:26652636 - 26761084 on Build GRCh38 | Insertion |
|
| esv3310588 | Chr.2:26714488 - 26714488 on Build GRCh38 | Novel Sequence Insertion |
|
| esv3370902 | Chr.2:26714489 - 26714489 on Build GRCh38 | Insertion |
|
| esv989007 | Chr.2:26714469 - 26716216 on Build GRCh38 | Insertion |
|
| nsv2652 | Chr.2:26696612 - 26723739 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map