This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03558914_cn
See other DHFR CNV Assays ›
Gene Symbol
DHFR
Assay Reference Genome
Location

Chr.5:80629671 on build GRCh38
Cytoband
5q14.1
Assay ID Hs03558914_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

1719

Gene Name:

dihydrofolate reductase

Gene Aliases:

DHFRP1, DYR

Location:

Chr.5:80626226-80654981 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DHFR NM_000791.3 NP_000782.1
NM_001290354.1 NP_001277283.1
NM_001290357.1 NP_001277286.1
NR_110936.1
AK293146.1
AK297302.1
AK308492.1
BC000192.1 AAH00192.1
BC003584.1 AAH03584.2
BC070280.1 AAH70280.1
BC071996.1 AAH71996.1
J00140.1 AAA58485.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv483112 Chr.5:80551019 - 80731514 on Build GRCh38 Loss MTRNR2L2 DHFR ANKRD34B MSH3 LINC01337
nsv4895 Chr.5:80623591 - 80640149 on Build GRCh38 Insertion DHFR

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
glycine biosynthetic process
tetrahydrobiopterin biosynthetic process
one-carbon metabolic process
nucleotide biosynthetic process
axon regeneration
response to methotrexate
dihydrofolate metabolic process
tetrahydrofolate metabolic process
tetrahydrofolate biosynthetic process
folic acid metabolic process
positive regulation of nitric-oxide synthase activity
oxidation-reduction process
regulation of removal of superoxide radicals
mRNA binding
dihydrofolate reductase activity
folic acid binding
drug binding
folate reductase activity
methotrexate binding
NADPH binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline