This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03670665_cn
See other PI15 CNV Assays ›
Gene Symbol
PI15
Assay Reference Genome
Location

Chr.8:74828667 on build GRCh38
Cytoband
8q21.13
Assay ID Hs03670665_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51050

Gene Name:

peptidase inhibitor 15

Gene Aliases:

CRISP8, P24TI, P25TI

Location:

Chr.8:74824537-74855044 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PI15 NM_001324403.1 NP_001311332.1
NM_015886.4 NP_056970.1
BC074931.2 AAH74931.1
BC074932.2 AAH74932.1
BC126290.1
BC126292.1
CR749657.1 CAH18451.2
D45027.1 BAA25066.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3891410 Chr.8:74600899 - 74942290 on Build GRCh38 Gain MIR2052 PI15 MIR2052HG
dgv7241n100 Chr.8:74778792 - 74838173 on Build GRCh38 Loss PI15
esv28358 Chr.8:74663344 - 75012983 on Build GRCh38 Gain+Loss MIR2052 PI15 CRISPLD1 MIR2052HG

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
negative regulation of peptidase activity
peptidase inhibitor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0