This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03726941_cn
See other LOC102724719 CNV Assays ›
Gene Symbol
LOC102724719
Assay Reference Genome
Location

Chr.10:51065166 on build GRCh38
Cytoband
10q11.23
Assay ID Hs03726941_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

102724719

Gene Name:

uncharacterized LOC102724719

Gene Aliases:

-

Location:

Chr.10:51062579-51068552 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC102724719 NR_120678.1
BF448553.1

Target Gene Details

Entrez Gene ID:

5592

Gene Name:

protein kinase, cGMP-dependent, type I

Gene Aliases:

AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha

Location:

Chr.10:50991151-52298350 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PRKG1 NM_001098512.2 NP_001091982.1
AK300110.1
D45864.1 BAA08297.1
DC310312.1
DC379897.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831873 Chr.10:50950850 - 51139780 on Build GRCh38 Gain LOC102724719 PRKG1
nsv1036645 Chr.10:51045201 - 51399316 on Build GRCh38 Gain MIR605 LOC102724719 PRKG1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
protein phosphorylation
signal transduction
dendrite development
cGMP-mediated signaling
actin cytoskeleton organization
forebrain development
regulation of GTPase activity
relaxation of vascular smooth muscle
negative regulation of platelet aggregation
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
cGMP-dependent protein kinase activity
calcium channel regulator activity
protein binding
ATP binding
cGMP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline