This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03749592_cn
See other DHTKD1 CNV Assays ›
Gene Symbol
DHTKD1
Assay Reference Genome
Location

Chr.10:12076088 on build GRCh38
Cytoband
10p14
Assay ID Hs03749592_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

55526

Gene Name:

dehydrogenase E1 and transketolase domain containing 1

Gene Aliases:

AMOXAD, CMT2Q

Location:

Chr.10:12068917-12123228 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DHTKD1 NM_018706.6 NP_061176.3
AK309810.1
BC002477.2 AAH02477.1
BC007955.2 AAH07955.1
CF593518.1
HY039281.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n21 Chr.10:12024028 - 12123248 on Build GRCh38 Gain DHTKD1 UPF2
esv3622354 Chr.10:12050805 - 12129371 on Build GRCh38 Gain DHTKD1
nsv1035894 Chr.10:12039222 - 12127501 on Build GRCh38 Gain DHTKD1 UPF2
esv3622357 Chr.10:12069904 - 12082451 on Build GRCh38 Gain DHTKD1
nsv1035709 Chr.10:12054410 - 12183299 on Build GRCh38 Gain MIR548AK DHTKD1 NUDT5 SEC61A2
nsv470919 Chr.10:12058736 - 12123248 on Build GRCh38 Gain DHTKD1
nsv831785 Chr.10:11928851 - 12124076 on Build GRCh38 Gain DHTKD1 UPF2
dgv673n100 Chr.10:12049505 - 12137652 on Build GRCh38 Gain MIR548AK DHTKD1 SEC61A2
nsv1046396 Chr.10:12059681 - 12109949 on Build GRCh38 Gain DHTKD1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
generation of precursor metabolites and energy
glycolytic process
tricarboxylic acid cycle
glyoxylate metabolic process
oxoglutarate dehydrogenase (succinyl-transferring) activity
thiamine pyrophosphate binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline