This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03770906_cn
See other CD44 CNV Assays ›
Gene Symbol
CD44
Assay Reference Genome
Location

Chr.11:35148347 on build GRCh38
Cytoband
11p13
Assay ID Hs03770906_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

960

Gene Name:

CD44 molecule (Indian blood group)

Gene Aliases:

CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1

Location:

Chr.11:35138870-35232402 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CD44 NM_000610.3 NP_000601.3
NM_001001389.1 NP_001001389.1
NM_001001390.1 NP_001001390.1
NM_001001391.1 NP_001001391.1
NM_001001392.1 NP_001001392.1
NM_001202555.1 NP_001189484.1
NM_001202556.1 NP_001189485.1
NM_001202557.1 NP_001189486.1
XM_005253231.2 XP_005253288.1
XM_005253232.2 XP_005253289.1
XM_005253233.2 XP_005253290.1
XM_005253234.2 XP_005253291.1
XM_005253235.2 XP_005253292.1
XM_005253238.2 XP_005253295.1
XM_005253239.2 XP_005253296.1
XM_005253240.2 XP_005253297.1
XM_006718388.1 XP_006718451.1
XM_006718390.3 XP_006718453.1
XM_011520482.1 XP_011518784.1
XM_011520483.1 XP_011518785.1
XM_011520484.1 XP_011518786.1
XM_011520485.1 XP_011518787.1
XM_011520486.1 XP_011518788.1
XM_011520487.2 XP_011518789.1
XM_011520488.1 XP_011518790.1
XM_011520489.2 XP_011518791.1
XM_017018583.1 XP_016874072.1
XM_017018584.1 XP_016874073.1
XM_017018585.1 XP_016874074.1
AF098641.1 AAC70782.1
AJ251595.1 CAB61878.1
AK290424.1
AK297776.1
AL832642.2 CAD89965.1
AY101192.1 AAM50040.1
AY101193.1 AAM50041.1
BC004372.1 AAH04372.1
BC052287.1 AAH52287.1
BC067348.1 AAH67348.1
EF581837.1
FJ216964.1
M24915.1 AAA35674.1
M25078.1 AAA36138.1
M59040.1 AAA51950.1
S66400.1 AAB27917.1
U40373.1 AAA82949.1
X55150.1 CAA38951.1
X56794.1 CAA40133.1
X66733.1 CAA47271.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2759818 Chr.11:35008386 - 35217031 on Build GRCh38 Loss LOC100507144 CD44
nsv1042014 Chr.11:34919190 - 37195907 on Build GRCh38 Loss TRAF6 LOC100507144 RAG1 PRR5L MIR3973 RAG2 COMMD9 LOC101928510 FJX1 C11orf74 PAMR1 CD44 MIR1343 SLC1A2 PDHX TRIM44 LDLRAD3
esv3625922 Chr.11:35148319 - 35190916 on Build GRCh38 Loss CD44
nsv508623 Chr.11:35073386 - 35181816 on Build GRCh38 Deletion LOC100507144 CD44

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell-matrix adhesion
single organismal cell-cell adhesion
extracellular matrix disassembly
extracellular matrix organization
hyaluronan catabolic process
positive regulation of peptidyl-serine phosphorylation
positive regulation of heterotypic cell-cell adhesion
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of DNA damage response, signal transduction by p53 class mediator
cellular response to fibroblast growth factor stimulus
positive regulation of peptidyl-tyrosine phosphorylation
leukocyte migration
cartilage development
interferon-gamma-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
monocyte aggregation
positive regulation of monocyte aggregation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
cytokine receptor activity
protein binding
collagen binding
hyaluronic acid binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline