This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03773155_cn
See other RASGRP2 CNV Assays ›
Gene Symbol
RASGRP2
Assay Reference Genome
Location

Chr.11:64730569 on build GRCh38
Cytoband
11q13.1
Assay ID Hs03773155_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10235

Gene Name:

RAS guanyl releasing protein 2

Gene Aliases:

CALDAG-GEFI, CDC25L

Location:

Chr.11:64726911-64745456 on Build GRCh38

Assay Gene Location:

Within Intron 16
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RASGRP2 NM_001098670.1 NP_001092140.1
NM_001098671.1 NP_001092141.1
NM_001318398.1 NP_001305327.1
NM_153819.1 NP_722541.1
XM_005273707.3 XP_005273764.3
XM_011544718.1 XP_011543020.1
XM_011544720.1 XP_011543022.1
XM_011544721.1 XP_011543023.1
XM_011544722.1 XP_011543024.1
XM_011544723.2 XP_011543025.1
XM_011544725.2 XP_011543027.1
XM_017017082.1 XP_016872571.1
XM_017017083.1 XP_016872572.1
XM_017017084.1 XP_016872573.1
XM_017017085.1 XP_016872574.1
XM_017017086.1 XP_016872575.1
AF043722.1 AAF07219.1
AF043723.1 AAF07220.1
AF081194.1 AAC79698.1
AK092852.1
AK092882.1
AK122694.1
AK314637.1
BC110306.1 AAI10307.1
BC117151.1
U78170.1 AAD12741.1
Y12336.1 CAA73005.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv818842 Chr.11:64577976 - 64778919 on Build GRCh38 Gain SF1 PYGM NRXN2 RASGRP2 SLC22A12
nsv359 Chr.11:64709536 - 64735401 on Build GRCh38 Deletion NRXN2 RASGRP2
dgv1965n54 Chr.11:64566642 - 64806117 on Build GRCh38 Loss SF1 SLC22A11 PYGM NRXN2 MAP4K2 RASGRP2 SLC22A12 MEN1
dgv1967n54 Chr.11:64709578 - 64817487 on Build GRCh38 Loss SF1 PYGM NRXN2 MAP4K2 RASGRP2 MEN1
nsv832189 Chr.11:64719711 - 64852727 on Build GRCh38 Loss EHD1 SF1 CDC42BPG PYGM NRXN2 MAP4K2 RASGRP2 MEN1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
signal transduction
Ras protein signal transduction
positive regulation of GTPase activity
cellular response to calcium ion
guanyl-nucleotide exchange factor activity
calcium ion binding
lipid binding
diacylglycerol binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline