This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03832927_cn
See other ACSM4 CNV Assays ›
Gene Symbol
ACSM4
Assay Reference Genome
Location

Chr.12:7308571 on build GRCh38
Cytoband
12p13.31
Assay ID Hs03832927_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

341392

Gene Name:

acyl-CoA synthetase medium-chain family member 4

Gene Aliases:

-

Location:

Chr.12:7304332-7328373 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ACSM4 NM_001080454.1 NP_001073923.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv448e212 Chr.12:7280402 - 7344654 on Build GRCh38 Gain ACSM4 CD163L1
nsv832325 Chr.12:7141408 - 7316916 on Build GRCh38 Gain+Loss CLSTN3 ACSM4 PEX5
dgv1349n100 Chr.12:7274380 - 7336757 on Build GRCh38 Gain ACSM4 CD163L1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
acyl-CoA metabolic process
fatty-acyl-CoA synthase activity
ATP binding
metal ion binding
butyrate-CoA ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline