This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03833562_cn
See other DDX11-AS1 CNV Assays ›
Gene Symbol
DDX11-AS1
Assay Reference Genome
Location

Chr.12:31039783 on build GRCh38
Cytoband
12p11.21
Assay ID Hs03833562_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100506660

Gene Name:

DDX11 antisense RNA 1

Gene Aliases:

-

Location:

Chr.12:31020763-31073847 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DDX11-AS1 NR_038927.2
AK096360.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv61e55 Chr.12:31018094 - 31261539 on Build GRCh38 Gain OVOS2 DDX11-AS1 DDX11
esv2751046 Chr.12:31021299 - 31531799 on Build GRCh38 Gain FAM60A OVOS2 DENND5B DDX11-AS1 FLJ13224 DDX11 LOC101929027
esv2422447 Chr.12:30801877 - 31296881 on Build GRCh38 Duplication FAM60A LINC00941 OVOS2 TSPAN11 DDX11-AS1 DDX11
nsv1038745 Chr.12:30934823 - 31152256 on Build GRCh38 Gain OVOS2 TSPAN11 DDX11-AS1 DDX11
esv3629033 Chr.12:31038557 - 31040199 on Build GRCh38 Loss DDX11-AS1
dgv7e196 Chr.12:30919025 - 31193061 on Build GRCh38 Duplication OVOS2 TSPAN11 DDX11-AS1 DDX11
esv3581161 Chr.12:30802222 - 31094869 on Build GRCh38 Gain LINC00941 TSPAN11 DDX11-AS1 DDX11
nsv558030 Chr.12:30816741 - 31113353 on Build GRCh38 Gain TSPAN11 DDX11-AS1 DDX11
nsv428276 Chr.12:30972532 - 31338170 on Build GRCh38 Gain FAM60A OVOS2 TSPAN11 DDX11-AS1 FLJ13224 DDX11
esv2658025 Chr.12:31038571 - 31040212 on Build GRCh38 Deletion DDX11-AS1
esv2751043 Chr.12:30995727 - 31346737 on Build GRCh38 Gain FAM60A OVOS2 TSPAN11 DDX11-AS1 FLJ13224 DDX11

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-dd86746ff-t8vjm:80/100.66.78.50:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline