This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03913107_cn
See other CPEB1 CNV Assays ›
Gene Symbol
CPEB1
Assay Reference Genome
Location

Chr.15:82560244 on build GRCh38
Cytoband
15q25.2
Assay ID Hs03913107_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

64506

Gene Name:

cytoplasmic polyadenylation element binding protein 1

Gene Aliases:

CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1

Location:

Chr.15:82543201-82648016 on Build GRCh38

Assay Gene Location:

Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPEB1 NM_001079533.1 NP_001073001.1
NM_001079534.1 NP_001073002.1
NM_001079535.1 NP_001073003.1
NM_001288819.1 NP_001275748.1
NM_001288820.1 NP_001275749.1
NM_030594.4 NP_085097.3
AF329402.1 AAK01239.1
AF329403.1 AAK01240.1
AK023265.1 BAB14496.1
AK294280.1
AK300105.1
AK301728.1
BC035348.1 AAH35348.1
BC050629.1 AAH50629.1
BX649159.1
DA340952.1
DA705313.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv483125 Chr.15:82448179 - 82617091 on Build GRCh38 Loss CPEB1 GOLGA6L17P RPS17 GOLGA2P10
nsv833071 Chr.15:82454948 - 82641759 on Build GRCh38 Gain CPEB1 GOLGA6L17P RPS17 GOLGA2P10
nsv1035899 Chr.15:82439721 - 82646980 on Build GRCh38 Gain LOC102724034 CPEB1 GOLGA6L17P RPS17 GOLGA2P10

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
translation
negative regulation of cell proliferation
positive regulation of neuron projection development
positive regulation of cell migration
cellular response to insulin stimulus
positive regulation of translation
mRNA transport
positive regulation of nitric-oxide synthase biosynthetic process
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to hypoxia
negative regulation of cytoplasmic translation
nucleotide binding
translation repressor activity, nucleic acid binding
mRNA 3'-UTR binding
translation factor activity, RNA binding
mRNA 3'-UTR AU-rich region binding
ribosome binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0