This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03954723_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5194444 on build GRCh38
Cytoband
17p13.2
Assay ID Hs03954723_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1
AK021878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain LOC400568 MED11 INCA1 LOC102724009 SLC52A1 NUP88 RABEP1 CAMTA2 CXCL16 LOC100130950 RPAIN ARRB2 KIF1C ZNF232 C17orf107 RNF167 PSMB6 SPAG7 ZFP3 MINK1 CHRNE PLD2 LOC101928000 GP1BA MIR6865 ZNF594 LOC101559451 ENO3 LOC101927979 C1QBP SMTNL2 USP6 ZMYND15 PFN1 SCIMP LOC107984973 VMO1 GLTPD2 PELP1 SLC25A11 ALOX15 TM4SF5 MIR6864 DHX33
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain ZNF594 LOC100130950 SCIMP RABEP1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0