This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03957027_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5206811 on build GRCh38
Cytoband
17p13.2
Assay ID Hs03957027_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1
AK021878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain MED11 ALOX15 DHX33 SLC25A11 INCA1 GLTPD2 CHRNE PFN1 ZNF232 ARRB2 PSMB6 LOC102724009 LOC101927979 RABEP1 VMO1 ZFP3 MIR6864 TM4SF5 LOC107984973 MINK1 CAMTA2 NUP88 C17orf107 SCIMP SLC52A1 LOC400568 SPAG7 CXCL16 LOC100130950 SMTNL2 RPAIN PLD2 USP6 ZMYND15 ZNF594 C1QBP MIR6865 GP1BA KIF1C PELP1 LOC101928000 RNF167 LOC101559451 ENO3
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain ZNF594 SCIMP LOC100130950 RABEP1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0