This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03957676_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5200463 on build GRCh38
Cytoband
17p13.2
Assay ID Hs03957676_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1
AK021878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain LOC100130950 MED11 LOC101559451 LOC101927979 LOC101928000 ENO3 CAMTA2 ZNF594 SCIMP CHRNE PFN1 GLTPD2 GP1BA PLD2 ZMYND15 SMTNL2 VMO1 RABEP1 PSMB6 KIF1C DHX33 SPAG7 C17orf107 MINK1 MIR6865 TM4SF5 RNF167 LOC107984973 INCA1 ARRB2 CXCL16 SLC25A11 LOC102724009 ZNF232 NUP88 C1QBP PELP1 MIR6864 ALOX15 SLC52A1 RPAIN USP6 ZFP3 LOC400568
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain LOC100130950 RABEP1 ZNF594 SCIMP

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0