This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03959424_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5197679 on build GRCh38
Cytoband
17p13.2
Assay ID Hs03959424_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1
AK021878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain MIR6864 SLC25A11 C1QBP LOC100130950 ALOX15 CHRNE CAMTA2 PSMB6 RPAIN RNF167 SLC52A1 VMO1 ZNF594 USP6 MINK1 PELP1 INCA1 GLTPD2 ARRB2 ENO3 ZNF232 DHX33 LOC400568 SMTNL2 LOC102724009 PLD2 MIR6865 NUP88 SPAG7 SCIMP LOC107984973 KIF1C ZMYND15 RABEP1 MED11 LOC101927979 TM4SF5 GP1BA CXCL16 LOC101928000 PFN1 C17orf107 ZFP3 LOC101559451
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain SCIMP LOC100130950 RABEP1 ZNF594

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline