This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03971920_cn
See other CD300C CNV Assays ›
Gene Symbol
CD300C
Assay Reference Genome
Location

Chr.17:74544237 on build GRCh38
Cytoband
17q25.1
Assay ID Hs03971920_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10871

Gene Name:

CD300c molecule

Gene Aliases:

CLM-6, CMRF-35, CMRF-35A, CMRF35, CMRF35-A1, CMRF35A, CMRF35A1, IGSF16, LIR

Location:

Chr.17:74534359-74546171 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CD300C NM_006678.4 NP_006669.1
XM_017024033.1 XP_016879522.1
XM_017024034.1 XP_016879523.1
BC022279.1 AAH22279.1
X66171.1 CAA46948.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv12e196 Chr.17:73675043 - 74565539 on Build GRCh38 Duplication BTBD17 CD300C KIF19 TTYH2 LOC400620 RPL38 GPR142 LOC100134391 CD300A DNAI2 LOC100130520 LINC00469 MGC16275 CD300LB GPRC5C
nsv470600 Chr.17:74466882 - 74550979 on Build GRCh38 Loss CD300A CD300C CD300LB
esv2761978 Chr.17:73835799 - 74684915 on Build GRCh38 Gain BTBD17 CD300C KIF19 TTYH2 RPL38 CD300E GPR142 C17orf77 CD300A DNAI2 LOC100130520 MGC16275 RAB37 LOC101928343 CD300LB GPRC5C CD300LD
esv3641230 Chr.17:73831787 - 74694861 on Build GRCh38 Gain BTBD17 CD300LF CD300C KIF19 TTYH2 RPL38 CD300E GPR142 C17orf77 CD300A DNAI2 LOC100130520 MGC16275 RAB37 LOC101928343 CD300LB GPRC5C CD300LD
dgv3282n100 Chr.17:73814343 - 74719597 on Build GRCh38 Gain BTBD17 CD300LF CD300C KIF19 TTYH2 RPL38 CD300E GPR142 C17orf77 CD300A DNAI2 LOC100130520 LINC00469 MGC16275 RAB37 LOC101928343 CD300LB GPRC5C CD300LD
esv33995 Chr.17:74261053 - 74732093 on Build GRCh38 Loss BTBD17 CD300LF CD300C KIF19 TTYH2 CD300E GPR142 C17orf77 CD300A DNAI2 LOC100130520 RAB37 LOC101928343 CD300LB GPRC5C CD300LD
dgv372n27 Chr.17:73826016 - 74685802 on Build GRCh38 Gain BTBD17 CD300C KIF19 TTYH2 RPL38 CD300E GPR142 C17orf77 CD300A DNAI2 LOC100130520 LINC00469 MGC16275 RAB37 LOC101928343 CD300LB GPRC5C CD300LD
esv2751770 Chr.17:73837862 - 74716223 on Build GRCh38 Gain BTBD17 CD300LF CD300C KIF19 TTYH2 RPL38 CD300E GPR142 C17orf77 CD300A DNAI2 LOC100130520 MGC16275 RAB37 LOC101928343 CD300LB GPRC5C CD300LD
esv3573820 Chr.17:73817932 - 74694378 on Build GRCh38 Gain BTBD17 CD300LF CD300C KIF19 TTYH2 RPL38 CD300E GPR142 C17orf77 CD300A DNAI2 LOC100130520 LINC00469 MGC16275 RAB37 LOC101928343 CD300LB GPRC5C CD300LD
dgv5774n54 Chr.17:74428381 - 74673021 on Build GRCh38 Gain C17orf77 CD300A LOC100130520 CD300C RAB37 CD300E LOC101928343 CD300LB GPRC5C CD300LD
esv3641244 Chr.17:74527395 - 74686883 on Build GRCh38 Gain C17orf77 LOC100130520 CD300C RAB37 CD300E LOC101928343 CD300LB CD300LD

Back To Top

More Information


Additional Information:

For this assay, SNP(s) [rs146645561] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cellular defense response
signal transduction
regulation of immune response
transmembrane signaling receptor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline