This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03976312_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5201864 on build GRCh38
Cytoband
17p13.2
Assay ID Hs03976312_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1
AK021878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain PELP1 LOC100130950 ZNF232 RNF167 ALOX15 TM4SF5 RABEP1 SLC25A11 KIF1C SCIMP LOC107984973 MIR6864 GP1BA LOC101927979 NUP88 LOC102724009 PSMB6 SPAG7 SLC52A1 MINK1 MIR6865 C1QBP USP6 INCA1 LOC400568 ZFP3 ARRB2 LOC101928000 CAMTA2 SMTNL2 GLTPD2 C17orf107 LOC101559451 VMO1 ENO3 MED11 RPAIN ZNF594 PLD2 CHRNE PFN1 CXCL16 DHX33 ZMYND15
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain LOC100130950 ZNF594 RABEP1 SCIMP

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline