This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03979416_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5208281 on build GRCh38
Cytoband
17p13.2
Assay ID Hs03979416_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1
AK021878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain MINK1 PLD2 C17orf107 PFN1 VMO1 MIR6865 LOC107984973 KIF1C RABEP1 ARRB2 DHX33 SLC52A1 ZNF232 MED11 ALOX15 C1QBP TM4SF5 LOC100130950 ZNF594 ZMYND15 RPAIN CHRNE ENO3 INCA1 GP1BA PELP1 CXCL16 LOC101928000 SPAG7 PSMB6 USP6 SLC25A11 SMTNL2 CAMTA2 LOC102724009 NUP88 LOC400568 ZFP3 LOC101927979 GLTPD2 SCIMP MIR6864 RNF167 LOC101559451
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain LOC100130950 ZNF594 SCIMP RABEP1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0