This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03979933_cn
See other CD300C CNV Assays ›
Gene Symbol
CD300C
Assay Reference Genome
Location

Chr.17:74544529 on build GRCh38
Cytoband
17q25.1
Assay ID Hs03979933_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10871

Gene Name:

CD300c molecule

Gene Aliases:

CLM-6, CMRF-35, CMRF-35A, CMRF35, CMRF35-A1, CMRF35A, CMRF35A1, IGSF16, LIR

Location:

Chr.17:74534359-74546171 on Build GRCh38

Assay Gene Location:

Overlaps Intron 2 - Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CD300C NM_006678.4 NP_006669.1
XM_017024033.1 XP_016879522.1
XM_017024034.1 XP_016879523.1
BC022279.1 AAH22279.1
X66171.1 CAA46948.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv12e196 Chr.17:73675043 - 74565539 on Build GRCh38 Duplication CD300C LINC00469 DNAI2 BTBD17 MGC16275 TTYH2 CD300LB CD300A KIF19 GPR142 GPRC5C LOC100130520 LOC100134391 LOC400620 RPL38
nsv470600 Chr.17:74466882 - 74550979 on Build GRCh38 Loss CD300C CD300LB CD300A
esv2761978 Chr.17:73835799 - 74684915 on Build GRCh38 Gain CD300LD CD300C DNAI2 BTBD17 MGC16275 TTYH2 CD300LB CD300A KIF19 CD300E GPR142 C17orf77 GPRC5C LOC100130520 LOC101928343 RAB37 RPL38
esv3641230 Chr.17:73831787 - 74694861 on Build GRCh38 Gain CD300LD CD300C DNAI2 BTBD17 MGC16275 TTYH2 CD300LB CD300A KIF19 CD300E GPR142 C17orf77 GPRC5C LOC100130520 LOC101928343 RAB37 RPL38 CD300LF
dgv3282n100 Chr.17:73814343 - 74719597 on Build GRCh38 Gain CD300LD CD300C LINC00469 DNAI2 BTBD17 MGC16275 TTYH2 CD300LB CD300A KIF19 CD300E GPR142 C17orf77 GPRC5C LOC100130520 LOC101928343 RAB37 RPL38 CD300LF
esv33995 Chr.17:74261053 - 74732093 on Build GRCh38 Loss CD300LD CD300C DNAI2 BTBD17 TTYH2 CD300LB CD300A KIF19 CD300E GPR142 C17orf77 GPRC5C LOC100130520 LOC101928343 RAB37 CD300LF
dgv372n27 Chr.17:73826016 - 74685802 on Build GRCh38 Gain CD300LD CD300C LINC00469 DNAI2 BTBD17 MGC16275 TTYH2 CD300LB CD300A KIF19 CD300E GPR142 C17orf77 GPRC5C LOC100130520 LOC101928343 RAB37 RPL38
esv2751770 Chr.17:73837862 - 74716223 on Build GRCh38 Gain CD300LD CD300C DNAI2 BTBD17 MGC16275 TTYH2 CD300LB CD300A KIF19 CD300E GPR142 C17orf77 GPRC5C LOC100130520 LOC101928343 RAB37 RPL38 CD300LF
esv3573820 Chr.17:73817932 - 74694378 on Build GRCh38 Gain CD300LD CD300C LINC00469 DNAI2 BTBD17 MGC16275 TTYH2 CD300LB CD300A KIF19 CD300E GPR142 C17orf77 GPRC5C LOC100130520 LOC101928343 RAB37 RPL38 CD300LF
dgv5774n54 Chr.17:74428381 - 74673021 on Build GRCh38 Gain CD300LD LOC101928343 CD300C RAB37 CD300LB CD300A CD300E C17orf77 GPRC5C LOC100130520
esv3641244 Chr.17:74527395 - 74686883 on Build GRCh38 Gain CD300LD LOC101928343 CD300C RAB37 CD300LB CD300E C17orf77 LOC100130520

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cellular defense response
signal transduction
regulation of immune response
transmembrane signaling receptor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0