This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03995282_cn
See other SOCS6 CNV Assays ›
Gene Symbol
SOCS6
Assay Reference Genome
Location

Chr.18:70302925 on build GRCh38
Cytoband
18q22.2
Assay ID Hs03995282_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9306

Gene Name:

suppressor of cytokine signaling 6

Gene Aliases:

CIS-4, CIS4, HSPC060, SOCS-4, SOCS-6, SOCS4, SSI4, STAI4, STATI4

Location:

Chr.18:70288901-70330199 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SOCS6 NM_004232.3 NP_004223.2
XM_005266783.3 XP_005266840.1
XM_017026087.1 XP_016881576.1
BC020082.1 AAH20082.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv32736 Chr.18:69135149 - 71678611 on Build GRCh38 Gain+Loss LINC01541 CCDC102B LOC101927481 SOCS6 LOC105372179 CD226 GTSCR1 LOC101060542 RTTN DOK6

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
defense response
JAK-STAT cascade
proteasomal protein catabolic process
protein ubiquitination
cytokine-mediated signaling pathway
regulation of growth
negative regulation of JAK-STAT cascade
negative regulation of T cell activation
protein kinase inhibitor activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-2x92s:80/100.66.79.168:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0