This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04013533_cn
See other ERCC2 CNV Assays ›
Gene Symbol
ERCC2
Assay Reference Genome
Location

Chr.19:45358498 on build GRCh38
Cytoband
19q13.32
Assay ID Hs04013533_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2068

Gene Name:

ERCC excision repair 2, TFIIH core complex helicase subunit

Gene Aliases:

COFS2, EM9, TFIIH, TTD, TTD1, XPD

Location:

Chr.19:45349837-45370647 on Build GRCh38

Assay Gene Location:

Within Intron 12
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ERCC2 NM_000400.3 NP_000391.1
XM_011526611.2 XP_011524913.1
XM_017026467.1 XP_016881956.1
AK092872.1 1 441
AK130849.1 BAC85446.1
BC108255.1 AAI08256.1
BC110522.1 AAI10523.1
BC110523.1 AAI10524.1
X52221.1 CAA36463.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv29n64 Chr.19:45334011 - 45373709 on Build GRCh38 Gain ERCC2 KLC3
nsv1160649 Chr.19:45353791 - 45361633 on Build GRCh38 Duplication ERCC2
nsv1063547 Chr.19:45322368 - 45400204 on Build GRCh38 Gain PPP1R13L CKM ERCC2 KLC3
nsv833844 Chr.19:45181051 - 45384148 on Build GRCh38 Loss PPP1R13L CKM ERCC2 BLOC1S3 MARK4 EXOC3L2 KLC3
dgv3606n100 Chr.19:45347435 - 45400204 on Build GRCh38 Gain PPP1R13L ERCC2 KLC3
nsv579868 Chr.19:45334025 - 45359625 on Build GRCh38 Gain ERCC2 KLC3
nsv442466 Chr.19:45347447 - 45388666 on Build GRCh38 Gain PPP1R13L ERCC2 KLC3
nsv428365 Chr.19:45358027 - 45469089 on Build GRCh38 Gain+Loss PPP1R13L FOSB CD3EAP ERCC2 MIR6088 ERCC1
nsv833845 Chr.19:45308814 - 45457371 on Build GRCh38 Loss PPP1R13L CKM CD3EAP ERCC2 MIR6088 ERCC1 KLC3
esv3644502 Chr.19:45323147 - 45403933 on Build GRCh38 Gain PPP1R13L ERCC2 KLC3
nsv515681 Chr.19:45352886 - 45364001 on Build GRCh38 Loss ERCC2
dgv1054e212 Chr.19:45347119 - 45407663 on Build GRCh38 Gain PPP1R13L CD3EAP ERCC2 ERCC1 KLC3
nsv579867 Chr.19:45323108 - 45408744 on Build GRCh38 Gain PPP1R13L CD3EAP ERCC2 ERCC1 KLC3
esv3644504 Chr.19:45342675 - 45373732 on Build GRCh38 Gain ERCC2 KLC3
nsv579869 Chr.19:45356868 - 45365051 on Build GRCh38 Loss ERCC2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
response to hypoxia
in utero embryonic development
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
protein phosphorylation
apoptotic process
response to oxidative stress
chromosome segregation
aging
cell proliferation
UV protection
post-embryonic development
viral process
spinal cord development
extracellular matrix organization
bone mineralization
central nervous system myelin formation
nucleotide-excision repair, DNA incision
multicellular organism growth
hair cell differentiation
embryonic cleavage
erythrocyte maturation
positive regulation of DNA binding
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
hair follicle maturation
hematopoietic stem cell differentiation
global genome nucleotide-excision repair
regulation of mitotic cell cycle phase transition
DNA binding
ATP-dependent DNA helicase activity
protein kinase activity
protein binding
ATP binding
protein C-terminus binding
DNA-dependent ATPase activity
RNA polymerase II carboxy-terminal domain kinase activity
5'-3' DNA helicase activity
metal ion binding
protein N-terminus binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0