This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04093096_cn
See other GSTT1 CNV Assays ›
Gene Symbol
GSTT1
Assay Reference Genome
Location

Chr.22_KI270879v1:277708 on build GRCh38
Cytoband
22_KI270879v1_alt
Assay ID Hs04093096_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2952

Gene Name:

glutathione S-transferase theta 1

Gene Aliases:

-

Location:

Chr.22_KI270879v1:270308-278486 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GSTT1 NM_000853.3 NP_000844.2
NM_001293807.1 NP_001280736.1
NM_001293808.1 NP_001280737.1
NM_001293809.1 NP_001280738.1
NM_001293810.1 NP_001280739.1
NM_001293811.1 NP_001280740.1
NM_001293812.1 NP_001280741.1
NM_001293813.1 NP_001280742.1
NM_001293814.1 NP_001280743.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv953056 Chr.22_KI270879v1:237452 - 291702 on Build GRCh38 Deletion GSTT1
esv3647426 Chr.22_KI270879v1:239419 - 285699 on Build GRCh38 Gain GSTT1
esv3575451 Chr.22_KI270879v1:248687 - 291702 on Build GRCh38 Gain GSTT1
dgv1302e212 Chr.22_KI270879v1:254597 - 291702 on Build GRCh38 Loss GSTT1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0