This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04105210_cn
See other GATA1 CNV Assays ›
Gene Symbol
GATA1
Assay Reference Genome
Location

Chr.X:48789577 on build GRCh38
Cytoband
Xp11.23
Assay ID Hs04105210_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2623

Gene Name:

GATA binding protein 1

Gene Aliases:

ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT

Location:

Chr.X:48786540-48794311 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GATA1 NM_002049.3 NP_002040.1
XM_011543897.2 XP_011542199.1
XM_011543898.2 XP_011542200.1
BC009797.1 AAH09797.1
M30601.1 AAA35885.1
X17254.1 CAA35120.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv528292 Chr.X:48433286 - 49274368 on Build GRCh38 Gain CCDC120 KCND1 HDAC6 FOXP3 PRICKLE3 PIM2 SLC35A2 TBC1D25 SYP GLOD5 LOC101927635 WDR45 TFE3 OTUD5 PPP1R3F PQBP1 WDR13 PRAF2 GPKOW SYP-AS1 WAS ERAS GRIPAP1 GATA1 PLP2 SUV39H1 CCDC22 RBM3 PCSK1N EBP FTSJ1 MAGIX TIMM17B SLC38A5 CACNA1F PORCN
esv32909 Chr.X:48003374 - 49285646 on Build GRCh38 Gain+Loss CCDC120 KCND1 SSX4B HDAC6 FOXP3 SSX9 ZNF630-AS1 PRICKLE3 PIM2 SLC35A2 SSX5 TBC1D25 SYP ZNF630 GLOD5 LOC101927635 WDR45 SSX1 TFE3 OTUD5 SSX3 SSX6 PPP1R3F PQBP1 WDR13 PRAF2 GPKOW SYP-AS1 WAS ERAS GRIPAP1 GATA1 PLP2 SSX4 SUV39H1 CCDC22 RBM3 SPACA5B PCSK1N EBP SPACA5 FTSJ1 MAGIX ZNF182 TIMM17B SLC38A5 CACNA1F PORCN
esv3576862 Chr.X:48755018 - 48837713 on Build GRCh38 Gain ERAS GLOD5 GATA1 HDAC6 PCSK1N
nsv6893 Chr.X:48778273 - 48810147 on Build GRCh38 Insertion GATA1 HDAC6

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
transcription from RNA polymerase II promoter
cell-cell signaling
heart development
blood coagulation
negative regulation of cell proliferation
male gonad development
organ morphogenesis
tissue development
regulation of glycoprotein biosynthetic process
regulation of definitive erythrocyte differentiation
regulation of primitive erythrocyte differentiation
erythrocyte differentiation
megakaryocyte differentiation
platelet formation
basophil differentiation
eosinophil differentiation
negative regulation of bone mineralization
positive regulation of osteoblast proliferation
embryonic hemopoiesis
eosinophil fate commitment
negative regulation of apoptotic process
positive regulation of erythrocyte differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
erythrocyte development
homeostasis of number of cells within a tissue
positive regulation of peptidyl-tyrosine phosphorylation
platelet aggregation
transcriptional activation by promoter-enhancer looping
dendritic cell differentiation
cellular response to thyroid hormone stimulus
negative regulation of transcription regulatory region DNA binding
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
p53 binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
DNA binding, bending
chromatin DNA binding
sequence-specific DNA binding
C2H2 zinc finger domain binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline