This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04111440_cn
See other RENBP CNV Assays ›
Gene Symbol
RENBP
Assay Reference Genome
Location

Chr.X:153936661 on build GRCh38
Cytoband
Xq28
Assay ID Hs04111440_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

5973

Gene Name:

renin binding protein

Gene Aliases:

RBP, RNBP

Location:

Chr.X:153935263-153944780 on Build GRCh38

Assay Gene Location:

Within Intron 9
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RENBP NM_002910.5 NP_002901.2
XM_017029698.1 XP_016885187.1
BC015558.2 AAH15558.1
D10232.1 BAA01082.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv33199 Chr.X:153065734 - 154591978 on Build GRCh38 Gain+Loss GDI1 ATP2B3 TREX2 G6PD LOC105373383 LOC105373377 IKBKG LOC107987332 CH17-340M24.3 NAA10 FAM58A LCA10 HCFC1-AS1 L1CAM MECP2 ARHGAP4 TKTL1 MIR3202-1 FAM3A OPN1LW EMD LOC105373381 LAGE3 LOC105373386 ABCD1 SRPK3 OPN1MW3 ZFP92 PDZD4 IRAK1 OPN1MW2 CTAG1A TMEM187 UBL4A PNCK BGN LOC105373378 TAZ FAM223A RPL10 DNASE1L1 PLXNA3 LOC107983954 RENBP OPN1MW FAM50A DUSP9 BCAP31 ATP6AP1 TEX28 SLC10A3 IDH3G HAUS7 MIR3202-2 FLNA PNMA6A SLC6A8 PLXNB3 MIR6858 ZNF275 SNORA70 HCFC1 MIR718 MAGEA1 SSR4 AVPR2
nsv469867 Chr.X:153813281 - 154018926 on Build GRCh38 Loss ARHGAP4 MIR3202-1 NAA10 MIR3202-2 LCA10 HCFC1-AS1 RENBP L1CAM HCFC1 PDZD4 IRAK1 TMEM187 AVPR2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

epimerase/racemase isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

N-acetylglucosamine metabolic process
UDP-N-acetylglucosamine biosynthetic process
N-acetylmannosamine metabolic process
regulation of blood pressure
negative regulation of endopeptidase activity
N-acetylneuraminate catabolic process
endopeptidase inhibitor activity
ATP binding
protein homodimerization activity
N-acylglucosamine 2-epimerase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0