This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04193582_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147208360 on build GRCh38
Cytoband
1q21.1
Assay ID Hs04193582_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 XP_005273003.1
XM_005272947.4 XP_005273004.1
XM_005272948.4 XP_005273005.1
XM_006711244.3 XP_006711307.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 XP_011507652.1
XM_011509351.2 XP_011507653.1
XM_017000801.1 XP_016856290.1
XM_017000802.1 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
AK314647.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain LOC728989 NBPF13P CH17-408M7.1 NBPF11 RNU1-13P PDZK1P1 NBPF12 RNVU1-8 LOC100132057 PDIA3P1 HYDIN2 FMO5 LOC284561 GJA8 NUDT4P2 RNVU1-7 LOC100996517 NBPF14 PDE4DIP NUDT4P1 RNVU1-6 MIR5087 ACP6 LOC100505824 PRKAB2 RNVU1-3 MIR6077 PPIAL4G NOTCH2NL LOC105371226 LOC105371242 RNVU1-1 LOC101928979 LOC100996763 LINC00624 GPR89B BCL9 LOC101927468 CHD1L NBPF10 LINC01138 GJA5
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss LOC728989 PDZK1 NBPF11 POLR3GL RNU1-13P RNVU1-8 LOC284561 NUDT4P2 RNVU1-7 NUDT4P1 MIR5087 ACP6 PRKAB2 MIR6077 PPIAL4G NOTCH2NL LOC105371226 RNVU1-1 LOC101928979 LOC100996763 LINC00624 ANKRD34A MIR6736 LOC101927468 RBM8A NBPF10 GNRHR2 LINC01138 GJA5 NBPF13P CH17-408M7.1 PDZK1P1 POLR3C NBPF12 LOC100132057 PDIA3P1 TXNIP HYDIN2 FMO5 GJA8 LOC100996517 NBPF14 PDE4DIP NUDT17 RNVU1-6 LOC100505824 LIX1L RNVU1-3 CD160 PEX11B LOC105371242 ITGA10 GPR89A GPR89B HFE2 RNF115 PIAS3 BCL9 ANKRD35 CHD1L
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss LOC728989 NBPF13P CH17-408M7.1 NBPF11 RNU1-13P PDZK1P1 NBPF12 RNVU1-8 LOC100132057 PDIA3P1 HYDIN2 FMO5 LOC284561 GJA8 NUDT4P2 RNVU1-7 LOC100996517 NBPF14 PDE4DIP NUDT4P1 MIR5087 ACP6 LOC100505824 PRKAB2 RNVU1-3 MIR6077 PPIAL4G NOTCH2NL LOC105371226 LOC105371242 RNVU1-1 LOC100996763 LINC00624 GPR89B BCL9 LOC101927468 CHD1L NBPF10 LINC01138 GJA5
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion LOC728989 NBPF13P CH17-408M7.1 NBPF11 POLR3GL RNU1-13P PDZK1P1 NBPF12 RNVU1-8 LOC100132057 PDIA3P1 TXNIP HYDIN2 FMO5 LOC284561 GJA8 NUDT4P2 RNVU1-7 LOC100996517 NBPF14 PDE4DIP NUDT4P1 RNVU1-6 MIR5087 ACP6 LOC100505824 PRKAB2 LIX1L RNVU1-3 MIR6077 PPIAL4G NOTCH2NL LOC105371226 LOC105371242 RNVU1-1 LOC101928979 LOC100996763 LINC00624 GPR89B HFE2 ANKRD34A BCL9 LOC101927468 CHD1L NBPF10 LINC01138 GJA5
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication LOC728989 NBPF13P CH17-408M7.1 NBPF11 RNU1-13P PDZK1P1 NBPF12 RNVU1-8 LOC100132057 PDIA3P1 HYDIN2 FMO5 LOC284561 GJA8 NUDT4P2 RNVU1-7 LOC100996517 NBPF14 PDE4DIP NUDT4P1 MIR5087 ACP6 LOC100505824 PRKAB2 RNVU1-3 MIR6077 PPIAL4G NOTCH2NL LOC105371226 LOC105371242 RNVU1-1 LOC100996763 LINC00624 GPR89B BCL9 LOC101927468 CHD1L LINC01138 GJA5
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain LOC728989 NBPF13P CH17-408M7.1 NBPF11 RNU1-13P PDZK1P1 NBPF12 RNVU1-8 LOC100132057 PDIA3P1 HYDIN2 FMO5 LOC284561 GJA8 NUDT4P2 RNVU1-7 LOC100996517 NBPF14 PDE4DIP NUDT4P1 RNVU1-6 MIR5087 ACP6 LOC100505824 PRKAB2 RNVU1-3 MIR6077 PPIAL4G NOTCH2NL LOC105371226 LOC105371242 RNVU1-1 LOC101928979 LOC100996763 LINC00624 GPR89B BCL9 LOC101927468 CHD1L NBPF10 LINC01138 GJA5
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion LOC728989 NBPF13P CH17-408M7.1 NBPF11 RNU1-13P PDZK1P1 NBPF12 RNVU1-8 LOC100132057 PDIA3P1 HYDIN2 FMO5 LOC284561 NBPF9 GJA8 NUDT4P2 RNVU1-7 LOC100996517 NBPF14 PDE4DIP NUDT4P1 MIR5087 ACP6 LOC100505824 PRKAB2 RNVU1-3 MIR6077 PPIAL4G NOTCH2NL LOC105371226 LOC105371242 RNVU1-1 LOC101928979 LOC100996763 LINC00624 GPR89B BCL9 LOC101927468 CHD1L NBPF10 LINC01138 GJA5
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss FMO5 PRKAB2 CHD1L PDIA3P1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline