This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04194169_cn
See other CDK11A CNV Assays ›
Gene Symbol
CDK11A
Assay Reference Genome
Location

Chr.1:1717287 on build GRCh38
Cytoband
1p36.33
Assay ID Hs04194169_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728642

Gene Name:

cyclin dependent kinase 11A

Gene Aliases:

CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA

Location:

Chr.1:1702383-1724565 on Build GRCh38

Assay Gene Location:

Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CDK11A NM_001313896.1 NP_001300825.1
NM_001313982.1 NP_001300911.1
NM_024011.3 NP_076916.2
NM_033529.3 NP_277071.2
NR_132739.1
NR_132740.1
AF067518.1 AAC72083.1
AF067519.1 AAC72084.1
AF067520.1 AAC72085.1
AF067521.1 AAC72086.1
AF067522.1 AAC72087.1
AF067523.1 AAC72088.1
AF067524.1 AAC72089.1
AF067526.1 AAC72091.1
AF067527.1 AAC72092.1
AF067528.1 AAC72093.1
AF067529.1 AAC72094.1
AK301495.1
AK303446.1
AK303451.1
BC028187.1
DC388505.1
U04819.1 AAA19585.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion FNDC10 CDK11A MIB2 SLC35E2B MMP23A SLC35E2 CDK11B GNB1 MMP23B NADK
dgv40n54 Chr.1:1706566 - 1749003 on Build GRCh38 Loss CDK11A SLC35E2
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss FNDC10 CDK11A MIB2 SLC35E2B MMP23A SLC35E2 SSU72 CDK11B MMP23B
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain CDK11A MIB2 SLC35E2B MMP23A CDK11B MMP23B
esv3585010 Chr.1:1670280 - 1751138 on Build GRCh38 Loss CDK11A SLC35E2B MMP23A SLC35E2
nsv436733 Chr.1:1717187 - 1717256 on Build GRCh38 Insertion CDK11A
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11A SLC35E2B MMP23A CDK11B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss FNDC10 CDK11A MIB2 SLC35E2B MMP23A SLC35E2 SSU72 CDK11B MMP23B NADK
nsv822475 Chr.1:1703561 - 1749146 on Build GRCh38 Gain CDK11A SLC35E2
dgv2e214 Chr.1:1695226 - 1749609 on Build GRCh38 Loss CDK11A MMP23A SLC35E2
dgv5e212 Chr.1:1710651 - 1756235 on Build GRCh38 Gain CDK11A SLC35E2 NADK
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain CDK11A SLC35E2B MMP23A SLC35E2 CDK11B MMP23B NADK
dgv39n54 Chr.1:1689422 - 1722233 on Build GRCh38 Loss CDK11A SLC35E2B MMP23A
dgv4e212 Chr.1:1696966 - 1756488 on Build GRCh38 Gain CDK11A MMP23A SLC35E2 NADK
nsv436343 Chr.1:1698179 - 1752741 on Build GRCh38 Deletion CDK11A MMP23A SLC35E2 NADK
nsv1152112 Chr.1:1717060 - 1718661 on Build GRCh38 Duplication CDK11A
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain CDK11A SLC35E2B MMP23A SLC35E2 CDK11B
nsv545031 Chr.1:1696548 - 1754601 on Build GRCh38 Gain CDK11A MMP23A SLC35E2 NADK
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss FNDC10 LOC105378949 GABRD MIB2 CFAP74 SLC35E2B SSU72 GNB1 ATAD3C MRPL20 VWA1 LOC107985729 CCNL2 CALML6 ANKRD65 LOC105378589 SLC35E2 ATAD3B PRKCZ LOC148413 NADK CDK11A TMEM88B CDK11B ATAD3A TMEM240 TMEM52 MMP23A LOC105378591 MMP23B LOC102724312
esv3577686 Chr.1:1685551 - 1721336 on Build GRCh38 Loss CDK11A SLC35E2B MMP23A
esv3585015 Chr.1:1696366 - 1742370 on Build GRCh38 Gain CDK11A MMP23A SLC35E2
nsv1011683 Chr.1:1696467 - 1756753 on Build GRCh38 Gain CDK11A MMP23A SLC35E2 NADK
nsv442858 Chr.1:1696479 - 1741164 on Build GRCh38 Gain+Loss CDK11A MMP23A SLC35E2
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss CDK11A MMP23A SLC35E2 GNB1 NADK
nsv822697 Chr.1:1703766 - 1735718 on Build GRCh38 Loss CDK11A SLC35E2
nsv545027 Chr.1:1688102 - 1732685 on Build GRCh38 Loss CDK11A SLC35E2B MMP23A SLC35E2
nsv822586 Chr.1:1703762 - 1719374 on Build GRCh38 Gain CDK11A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss PLEKHN1 LOC100506504 SLC35E2B SSU72 GNB1 LOC100129534 ATAD3C VWA1 PUSL1 MIR6859-1 LOC100130417 CDK11A LINC01128 TTLL10-AS1 CDK11B SCNN1D DDX11L1 LOC100132287 ATAD3A FAM132A RNF223 LOC105378591 TNFRSF18 LOC105378592 HES4 LOC100133331 LOC105378949 GABRD MIB2 CPSF3L MIR1302-2 PRKCZ LOC148413 C1orf159 LOC105378947 OR4F29 LOC284600 MIR6808 LOC107984841 MIR429 LINC00115 WASH7P TMEM240 UBE2J2 LOC100287934 TTLL10 MIR6727 CFAP74 CPTP ISG15 CCNL2 CALML6 FAAP20 SLC35E2 ATAD3B NADK LOC100288175 MORN1 LOC100288069 MXRA8 OR4F5 TMEM88B TNFRSF4 SAMD11 AURKAIP1 MTND1P23 LOC107985728 AGRN LINC01342 FNDC10 PERM1 SKI MIR6859-2 B3GALT6 LOC102725121 MRPL20 LOC100134822 NOC2L LOC107985729 MIR6726 FAM138A ANKRD65 LOC105378589 DVL1 TAS1R3 KLHL17 MIR6723 MTND2P28 FAM87B SDF4 LOC101928626 ACAP3 FAM41C LOC105378948 OR4F16 TMEM52 MMP23A MMP23B MIR200A MIR200B LOC729737 LOC102724312
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication FNDC10 CDK11A MIB2 SLC35E2B MMP23A SLC35E2 SSU72 CDK11B GNB1 MMP23B NADK
esv3567896 Chr.1:1716171 - 1741152 on Build GRCh38 Gain CDK11A SLC35E2
nsv822809 Chr.1:1716430 - 1735718 on Build GRCh38 Loss CDK11A SLC35E2
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss PLEKHN1 MIR6727 SLC35E2B CPTP SSU72 ATAD3C ISG15 VWA1 PUSL1 CCNL2 LOC100130417 SLC35E2 ATAD3B NADK LOC100288175 CDK11A LOC100288069 MXRA8 LINC01128 TTLL10-AS1 TMEM88B CDK11B SCNN1D TNFRSF4 ATAD3A FAM132A SAMD11 AURKAIP1 RNF223 TNFRSF18 LOC107985728 AGRN LINC01342 HES4 FNDC10 MIB2 CPSF3L PERM1 B3GALT6 MRPL20 NOC2L LOC107985729 MIR6726 ANKRD65 DVL1 LOC148413 C1orf159 TAS1R3 KLHL17 LOC284600 FAM87B MIR6808 SDF4 MIR429 LINC00115 ACAP3 FAM41C LOC105378948 TMEM240 UBE2J2 MMP23A LOC100287934 MMP23B TTLL10 MIR200A MIR200B LOC102724312
dgv41n54 Chr.1:1713469 - 1740343 on Build GRCh38 Loss CDK11A SLC35E2
nsv482168 Chr.1:1702731 - 1724352 on Build GRCh38 Gain CDK11A
nsv1160925 Chr.1:1696979 - 1756148 on Build GRCh38 Deletion CDK11A MMP23A SLC35E2 NADK

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
mitotic nuclear division
regulation of mRNA processing
regulation of cell cycle
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0