This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04194878_cn
See other SLC35E2B CNV Assays ›
Gene Symbol
SLC35E2B
Assay Reference Genome
Location

Chr.1:1672677 on build GRCh38
Cytoband
1p36.33
Assay ID Hs04194878_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728661

Gene Name:

solute carrier family 35 member E2B

Gene Aliases:

SLC35E2

Location:

Chr.1:1661478-1692804 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC35E2B NM_001110781.2 NP_001104251.1
NM_001290264.1 NP_001277193.1
XM_011542072.1 XP_011540374.1
AB007916.3
AK125619.1
AK307886.1
BC110653.1 AAI10654.1
CR936618.1 CAI56761.1
DA794179.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n54 Chr.1:1663759 - 1696548 on Build GRCh38 Loss SLC35E2B
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion CDK11A CDK11B MMP23B SLC35E2 NADK GNB1 FNDC10 SLC35E2B MIB2 MMP23A
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss CDK11A CDK11B MMP23B SLC35E2 SSU72 FNDC10 SLC35E2B MIB2 MMP23A
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain CDK11A CDK11B MMP23B SLC35E2B MIB2 MMP23A
esv3585010 Chr.1:1670280 - 1751138 on Build GRCh38 Loss CDK11A SLC35E2 SLC35E2B MMP23A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC105378592 LOC148413 SSU72 MIR6859-2 DDX11L1 LOC100288175 MIB2 LOC105378948 AGRN MIR6726 SLC35E2B CPSF3L MTND2P28 LOC101928626 LOC100132287 MTND1P23 SCNN1D LINC01342 MIR1302-2 NOC2L SAMD11 MMP23A VWA1 MIR6859-1 ATAD3B ATAD3A OR4F29 C1orf159 TNFRSF4 PRKCZ MIR200B MIR6723 SDF4 FAM132A LOC102725121 CCNL2 TMEM88B LOC105378949 CDK11B CFAP74 CALML6 WASH7P TAS1R3 TMEM240 MIR200A LOC729737 LOC105378947 LOC100134822 SKI LOC284600 MORN1 MIR429 FAM138A PUSL1 LOC100129534 LOC100287934 TTLL10-AS1 GABRD LOC107985728 TNFRSF18 PLEKHN1 TTLL10 TMEM52 LINC01128 LOC100130417 ATAD3C GNB1 FAM87B CPTP RNF223 LOC100133331 MMP23B B3GALT6 ACAP3 HES4 DVL1 LOC105378589 MXRA8 KLHL17 LINC00115 LOC107984841 OR4F5 MIR6727 AURKAIP1 LOC102724312 FAAP20 CDK11A ISG15 UBE2J2 LOC100506504 FNDC10 PERM1 MRPL20 MIR6808 SLC35E2 NADK LOC105378591 FAM41C OR4F16 LOC107985729 LOC100288069 ANKRD65
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication CDK11A CDK11B MMP23B SLC35E2 NADK GNB1 SSU72 FNDC10 SLC35E2B MIB2 MMP23A
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11A CDK11B SLC35E2B MMP23A
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss CDK11A CDK11B MMP23B SLC35E2 NADK SSU72 FNDC10 SLC35E2B MIB2 MMP23A
esv3585008 Chr.1:1663929 - 1681002 on Build GRCh38 Loss SLC35E2B
nsv945745 Chr.1:1671930 - 1674224 on Build GRCh38 Duplication SLC35E2B
esv3585007 Chr.1:1647187 - 1714507 on Build GRCh38 Loss CDK11A CDK11B SLC35E2B MMP23A
nsv950516 Chr.1:1663141 - 1715961 on Build GRCh38 Deletion CDK11A SLC35E2B MMP23A
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain CDK11A CDK11B MMP23B SLC35E2 NADK SLC35E2B MMP23A
nsv545022 Chr.1:1665581 - 1683909 on Build GRCh38 Loss SLC35E2B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC148413 LOC107985728 SSU72 TNFRSF18 PLEKHN1 TTLL10 LINC01128 LOC100130417 ATAD3C LOC100288175 FAM87B CPTP MIB2 RNF223 LOC105378948 MMP23B AGRN B3GALT6 ACAP3 MIR6726 SLC35E2B CPSF3L HES4 DVL1 MXRA8 KLHL17 SCNN1D LINC00115 LINC01342 NOC2L SAMD11 MMP23A MIR6727 VWA1 AURKAIP1 ATAD3B ATAD3A LOC102724312 C1orf159 CDK11A ISG15 UBE2J2 TNFRSF4 MIR200B FNDC10 PERM1 SDF4 MRPL20 FAM132A CCNL2 TMEM88B MIR6808 CDK11B SLC35E2 NADK TAS1R3 TMEM240 MIR200A FAM41C LOC107985729 LOC284600 MIR429 LOC100288069 ANKRD65 PUSL1 LOC100287934 TTLL10-AS1
nsv545026 Chr.1:1671348 - 1706566 on Build GRCh38 Loss CDK11A SLC35E2B MMP23A
nsv545018 Chr.1:1638499 - 1688102 on Build GRCh38 Loss CDK11B SLC35E2B
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain CDK11A CDK11B SLC35E2 SLC35E2B MMP23A
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss LOC148413 GABRD VWA1 SSU72 ATAD3B TMEM52 ATAD3A LOC102724312 CDK11A ATAD3C GNB1 PRKCZ FNDC10 MRPL20 MIB2 CCNL2 TMEM88B LOC105378949 CDK11B MMP23B SLC35E2 CFAP74 CALML6 NADK TMEM240 LOC105378591 SLC35E2B LOC107985729 LOC105378589 ANKRD65 MMP23A

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0