This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04198988_cn
See other CDK11A CNV Assays ›
Gene Symbol
CDK11A
Assay Reference Genome
Location

Chr.1:1721458 on build GRCh38
Cytoband
1p36.33
Assay ID Hs04198988_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728642

Gene Name:

cyclin dependent kinase 11A

Gene Aliases:

CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA

Location:

Chr.1:1702383-1724565 on Build GRCh38

Assay Gene Location:

Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CDK11A NM_001313896.1 NP_001300825.1
NM_001313982.1 NP_001300911.1
NM_024011.3 NP_076916.2
NM_033529.3 NP_277071.2
NR_132739.1
NR_132740.1
AF067518.1 AAC72083.1
AF067519.1 AAC72084.1
AF067520.1 AAC72085.1
AF067521.1 AAC72086.1
AF067522.1 AAC72087.1
AF067523.1 AAC72088.1
AF067526.1 AAC72091.1
AF067527.1 AAC72092.1
AF067528.1 AAC72093.1
AF067529.1 AAC72094.1
AK301495.1
AK303446.1
AK303451.1
BC028187.1
DC388505.1
U04819.1 AAA19585.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion CDK11A SLC35E2 CDK11B GNB1 MIB2 FNDC10 MMP23B MMP23A SLC35E2B NADK
dgv40n54 Chr.1:1706566 - 1749003 on Build GRCh38 Loss CDK11A SLC35E2
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss CDK11A SLC35E2 CDK11B SSU72 MIB2 FNDC10 MMP23B MMP23A SLC35E2B
esv3585010 Chr.1:1670280 - 1751138 on Build GRCh38 Loss CDK11A SLC35E2 MMP23A SLC35E2B
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11A CDK11B MMP23A SLC35E2B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss CDK11A SLC35E2 CDK11B SSU72 MIB2 FNDC10 MMP23B MMP23A SLC35E2B NADK
nsv822475 Chr.1:1703561 - 1749146 on Build GRCh38 Gain CDK11A SLC35E2
dgv2e214 Chr.1:1695226 - 1749609 on Build GRCh38 Loss CDK11A SLC35E2 MMP23A
dgv5e212 Chr.1:1710651 - 1756235 on Build GRCh38 Gain CDK11A SLC35E2 NADK
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain CDK11A SLC35E2 CDK11B MMP23B MMP23A SLC35E2B NADK
dgv39n54 Chr.1:1689422 - 1722233 on Build GRCh38 Loss CDK11A MMP23A SLC35E2B
dgv4e212 Chr.1:1696966 - 1756488 on Build GRCh38 Gain CDK11A SLC35E2 MMP23A NADK
nsv436343 Chr.1:1698179 - 1752741 on Build GRCh38 Deletion CDK11A SLC35E2 MMP23A NADK
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain CDK11A SLC35E2 CDK11B MMP23A SLC35E2B
nsv545031 Chr.1:1696548 - 1754601 on Build GRCh38 Gain CDK11A SLC35E2 MMP23A NADK
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss TMEM88B CDK11A ATAD3C CDK11B CALML6 GNB1 MRPL20 LOC102724312 PRKCZ NADK LOC105378591 ATAD3B TMEM240 MIB2 MMP23B SLC35E2B LOC105378949 ANKRD65 TMEM52 SLC35E2 LOC105378589 CCNL2 SSU72 GABRD VWA1 LOC107985729 MMP23A LOC148413 FNDC10 ATAD3A CFAP74
dgv3e212 Chr.1:1685551 - 1756235 on Build GRCh38 Loss CDK11A SLC35E2 MMP23A SLC35E2B NADK
esv3585015 Chr.1:1696366 - 1742370 on Build GRCh38 Gain CDK11A SLC35E2 MMP23A
nsv1011683 Chr.1:1696467 - 1756753 on Build GRCh38 Gain CDK11A SLC35E2 MMP23A NADK
nsv442858 Chr.1:1696479 - 1741164 on Build GRCh38 Gain+Loss CDK11A SLC35E2 MMP23A
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss CDK11A SLC35E2 GNB1 MMP23A NADK
nsv822697 Chr.1:1703766 - 1735718 on Build GRCh38 Loss CDK11A SLC35E2
nsv545027 Chr.1:1688102 - 1732685 on Build GRCh38 Loss CDK11A SLC35E2 MMP23A SLC35E2B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CDK11A ATAD3C CDK11B GNB1 LOC107985728 MORN1 NADK LOC100134822 WASH7P MIB2 MMP23B LOC105378592 LINC01128 ANKRD65 MIR1302-2 TTLL10 MIR6808 SSU72 MIR6727 VWA1 MIR200A LOC729737 OR4F29 PLEKHN1 MIR429 MIR6859-1 FAM138A MIR6723 FAAP20 DDX11L1 PUSL1 CALML6 LOC107984841 LOC100288175 LOC102724312 MTND1P23 UBE2J2 PERM1 LOC100288069 ATAD3B AGRN LOC105378947 LOC101928626 LOC105378589 FAM87B B3GALT6 GABRD LOC107985729 KLHL17 LINC00115 SDF4 ACAP3 CPSF3L LOC100133331 PRKCZ LOC105378591 RNF223 TMEM240 MXRA8 OR4F16 SLC35E2B OR4F5 ISG15 SLC35E2 LOC102725121 LOC105378948 MMP23A HES4 CPTP MTND2P28 MIR6726 LOC148413 FNDC10 LINC01342 FAM41C TMEM88B LOC100130417 MIR200B TAS1R3 SKI MRPL20 DVL1 NOC2L SCNN1D TTLL10-AS1 FAM132A LOC105378949 TMEM52 LOC100287934 LOC284600 CCNL2 MIR6859-2 TNFRSF18 TNFRSF4 AURKAIP1 SAMD11 LOC100132287 LOC100129534 C1orf159 LOC100506504 ATAD3A CFAP74
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication CDK11A SLC35E2 CDK11B GNB1 SSU72 MIB2 FNDC10 MMP23B MMP23A SLC35E2B NADK
esv3567896 Chr.1:1716171 - 1741152 on Build GRCh38 Gain CDK11A SLC35E2
nsv822809 Chr.1:1716430 - 1735718 on Build GRCh38 Loss CDK11A SLC35E2
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CDK11A ATAD3C CDK11B CPSF3L LOC107985728 NADK RNF223 TMEM240 MIB2 MMP23B LINC01128 MXRA8 SLC35E2B ISG15 ANKRD65 SLC35E2 TTLL10 MIR6808 SSU72 MIR6727 VWA1 MIR200A LOC105378948 MMP23A HES4 PLEKHN1 MIR429 CPTP MIR6726 LOC148413 FNDC10 LINC01342 FAM41C TMEM88B PUSL1 LOC100130417 MIR200B TAS1R3 MRPL20 DVL1 NOC2L LOC100288175 LOC102724312 SCNN1D UBE2J2 PERM1 LOC100288069 ATAD3B TTLL10-AS1 FAM132A AGRN LOC100287934 LOC284600 CCNL2 TNFRSF18 FAM87B B3GALT6 LOC107985729 KLHL17 TNFRSF4 AURKAIP1 SAMD11 LINC00115 C1orf159 SDF4 ACAP3 ATAD3A
dgv41n54 Chr.1:1713469 - 1740343 on Build GRCh38 Loss CDK11A SLC35E2
nsv482168 Chr.1:1702731 - 1724352 on Build GRCh38 Gain CDK11A
nsv1160925 Chr.1:1696979 - 1756148 on Build GRCh38 Deletion CDK11A SLC35E2 MMP23A NADK

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
mitotic nuclear division
regulation of mRNA processing
regulation of cell cycle
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline