This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04210219_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147185588 on build GRCh38
Cytoband
1q21.1
Assay ID Hs04210219_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
XM_005272946.4 9 3277 XP_005273003.1
XM_005272947.4 9 3021 XP_005273004.1
XM_005272948.4 9 3004 XP_005273005.1
XM_006711245.3 8 2940 XP_006711308.1
XM_011509350.2 9 3308 XP_011507652.1
XM_011509351.2 8 3040 XP_011507653.1
XM_017000801.1 8 3052 XP_016856290.1
XM_017000802.1 8 2822 XP_016856291.1
AK225739.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain NBPF12 ACP6 LOC100996763 LOC100996517 RNVU1-1 NBPF14 BCL9 LOC105371242 GJA8 CH17-408M7.1 PRKAB2 NUDT4P2 FMO5 RNU1-13P RNVU1-8 GJA5 LOC728989 LOC284561 PDIA3P1 NOTCH2NL LOC101927468 PDE4DIP RNVU1-7 RNVU1-3 CHD1L RNVU1-6 PPIAL4G LINC00624 NBPF10 LINC01138 LOC100132057 NBPF13P MIR6077 HYDIN2 LOC105371226 LOC101928979 PDZK1P1 NBPF11 GPR89B MIR5087 LOC100505824 NUDT4P1
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss NBPF12 LOC100996763 LOC100996517 PIAS3 RNVU1-1 NBPF14 LIX1L CH17-408M7.1 NUDT4P2 FMO5 RNU1-13P RNVU1-8 GJA5 LOC284561 ANKRD34A PDIA3P1 NOTCH2NL PDE4DIP RNVU1-7 CHD1L RNVU1-6 PPIAL4G GNRHR2 LOC100132057 MIR6077 HYDIN2 LOC105371226 LOC101928979 PDZK1P1 NBPF11 PEX11B GPR89B POLR3C ITGA10 LOC100505824 ACP6 TXNIP MIR6736 BCL9 LOC105371242 NUDT17 ANKRD35 GJA8 PRKAB2 LOC728989 LOC101927468 RNF115 RNVU1-3 GPR89A LINC00624 NBPF10 LINC01138 NBPF13P POLR3GL CD160 PDZK1 MIR5087 HFE2 NUDT4P1 RBM8A
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion FMO5 RNVU1-8 LOC728989 PDIA3P1 NBPF13P PRKAB2 CHD1L
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss NBPF12 ACP6 LOC100996763 LOC100996517 RNVU1-1 NBPF14 BCL9 LOC105371242 GJA8 CH17-408M7.1 PRKAB2 NUDT4P2 FMO5 RNU1-13P RNVU1-8 GJA5 LOC728989 LOC284561 PDIA3P1 NOTCH2NL LOC101927468 PDE4DIP RNVU1-7 RNVU1-3 CHD1L PPIAL4G LINC00624 NBPF10 LINC01138 LOC100132057 NBPF13P MIR6077 HYDIN2 LOC105371226 PDZK1P1 NBPF11 GPR89B MIR5087 LOC100505824 NUDT4P1
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion NBPF12 ACP6 TXNIP LOC100996763 LOC100996517 RNVU1-1 NBPF14 BCL9 LOC105371242 LIX1L GJA8 CH17-408M7.1 PRKAB2 NUDT4P2 FMO5 RNU1-13P RNVU1-8 GJA5 LOC728989 LOC284561 ANKRD34A PDIA3P1 NOTCH2NL LOC101927468 PDE4DIP RNVU1-7 RNVU1-3 CHD1L RNVU1-6 PPIAL4G LINC00624 NBPF10 LINC01138 LOC100132057 NBPF13P MIR6077 HYDIN2 LOC105371226 POLR3GL LOC101928979 PDZK1P1 NBPF11 GPR89B MIR5087 HFE2 LOC100505824 NUDT4P1
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication NBPF12 ACP6 LOC100996763 LOC100996517 RNVU1-1 NBPF14 BCL9 LOC105371242 GJA8 CH17-408M7.1 PRKAB2 NUDT4P2 FMO5 RNU1-13P RNVU1-8 GJA5 LOC728989 LOC284561 PDIA3P1 NOTCH2NL LOC101927468 PDE4DIP RNVU1-7 RNVU1-3 CHD1L PPIAL4G LINC00624 LINC01138 LOC100132057 NBPF13P MIR6077 HYDIN2 LOC105371226 PDZK1P1 NBPF11 GPR89B MIR5087 LOC100505824 NUDT4P1
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain NBPF12 ACP6 LOC100996763 LOC100996517 RNVU1-1 NBPF14 BCL9 LOC105371242 GJA8 CH17-408M7.1 PRKAB2 NUDT4P2 FMO5 RNU1-13P RNVU1-8 GJA5 LOC728989 LOC284561 PDIA3P1 NOTCH2NL LOC101927468 PDE4DIP RNVU1-7 RNVU1-3 CHD1L RNVU1-6 PPIAL4G LINC00624 NBPF10 LINC01138 LOC100132057 NBPF13P MIR6077 HYDIN2 LOC105371226 LOC101928979 PDZK1P1 NBPF11 GPR89B MIR5087 LOC100505824 NUDT4P1
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion NBPF12 ACP6 LOC100996763 LOC100996517 RNVU1-1 NBPF9 NBPF14 BCL9 LOC105371242 GJA8 CH17-408M7.1 PRKAB2 NUDT4P2 FMO5 RNU1-13P RNVU1-8 GJA5 LOC728989 LOC284561 PDIA3P1 NOTCH2NL LOC101927468 PDE4DIP RNVU1-7 RNVU1-3 CHD1L PPIAL4G LINC00624 NBPF10 LINC01138 LOC100132057 NBPF13P MIR6077 HYDIN2 LOC105371226 LOC101928979 PDZK1P1 NBPF11 GPR89B MIR5087 LOC100505824 NUDT4P1
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss FMO5 PDIA3P1 PRKAB2 CHD1L

Back To Top

More Information


Set Membership:

Intragenic Exonic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline