This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04226786_cn
See other SLC40A1 CNV Assays ›
Gene Symbol
SLC40A1
Assay Reference Genome
Location

Chr.2:189567765 on build GRCh38
Cytoband
2q32.2
Assay ID Hs04226786_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

30061

Gene Name:

solute carrier family 40 member 1

Gene Aliases:

FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3

Location:

Chr.2:189560590-189580811 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC40A1 NM_014585.5 NP_055400.1
XM_005246505.1 XP_005246562.1
XM_017003938.1 XP_016859427.1
AF215636.1 AAF80986.1
AF226614.1 AAF36697.1
AF231121.1 AAF44330.1
AK223236.1 BAD96956.1
AK292146.1
AK314827.1
AL136944.1 CAB66878.1
BC035893.1 AAH35893.1
BC037733.1 AAH37733.1
CR533564.1 CAG38595.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv954448 Chr.2:189550075 - 189580174 on Build GRCh38 Duplication SLC40A1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

lymphocyte homeostasis
endothelium development
cellular iron ion homeostasis
regulation of transcription from RNA polymerase II promoter in response to iron
iron ion transmembrane transport
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
spleen trabecula formation
multicellular organismal iron ion homeostasis
ferrous iron export
receptor activity
iron ion transmembrane transporter activity
protein binding
ferrous iron transmembrane transporter activity
peptide hormone binding
iron channel activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline