This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404518_cn
See other DDX11 CNV Assays ›
Gene Symbol
DDX11
Assay Reference Genome
Location

Chr.12:31097210 on build GRCh38
Cytoband
12p11.21
Assay ID Hs04404518_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

1663

Gene Name:

DEAD/H-box helicase 11

Gene Aliases:

CHL1, CHLR1, KRG2, WABS

Location:

Chr.12:31073574-31104799 on Build GRCh38

Assay Gene Location:

Within Intron 22
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DDX11 NM_001257144.1 NP_001244073.1
NM_001257145.1 NP_001244074.1
NM_004399.2 NP_004390.3
NM_030653.3 NP_085911.2
NM_152438.1 NP_689651.1
XM_005253331.3 XP_005253388.1
XM_005253333.3 XP_005253390.1
XM_006719050.3 XP_006719113.1
XM_011520592.2 XP_011518894.1
XM_011520595.2 XP_011518897.1
XM_011520597.2 XP_011518899.1
XM_017018918.1 XP_016874407.1
XM_017018919.1 XP_016874408.1
XM_017018920.1 XP_016874409.1
XM_017018921.1 XP_016874410.1
XM_017018922.1 XP_016874411.1
XM_017018923.1 XP_016874412.1
XM_017018924.1 XP_016874413.1
XM_017018925.1 XP_016874414.1
XM_017018926.1 XP_016874415.1
XM_017018927.1 XP_016874416.1
XM_017018928.1 XP_016874417.1
XM_017018929.1 XP_016874418.1
XM_017018930.1 XP_016874419.1
XM_017018931.1 XP_016874420.1
XM_017018932.1 XP_016874421.1
XM_017018933.1 XP_016874422.1
XM_017018934.1 XP_016874423.1
XM_017018935.1 XP_016874424.1
XM_017018936.1 XP_016874425.1
XM_017018937.1 XP_016874426.1
XM_017018938.1 XP_016874427.1
XM_017018939.1 XP_016874428.1
BC050069.1 AAH50069.1
BC050522.1 AAH50522.1
BC111733.1 AAI11734.1
U33833.1 AAB06962.1
U75967.1 AAB18749.1
U75968.1 AAB18750.1
U75969.1 AAB18751.1
X99583.1 CAA67895.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv61e55 Chr.12:31018094 - 31261539 on Build GRCh38 Gain OVOS2 DDX11 DDX11-AS1
esv2751046 Chr.12:31021299 - 31531799 on Build GRCh38 Gain OVOS2 DENND5B LOC101929027 FAM60A FLJ13224 DDX11 DDX11-AS1
nsv1038745 Chr.12:30934823 - 31152256 on Build GRCh38 Gain OVOS2 TSPAN11 DDX11 DDX11-AS1
esv2444127 Chr.12:31092569 - 31175366 on Build GRCh38 Loss OVOS2 DDX11
nsv509462 Chr.12:31094479 - 31128240 on Build GRCh38 Insertion OVOS2 DDX11
dgv8e196 Chr.12:31066013 - 31129217 on Build GRCh38 Duplication OVOS2 DDX11 DDX11-AS1
esv2751043 Chr.12:30995727 - 31346737 on Build GRCh38 Gain OVOS2 TSPAN11 FAM60A FLJ13224 DDX11 DDX11-AS1
nsv975470 Chr.12:31077366 - 31107608 on Build GRCh38 Duplication DDX11
esv2422283 Chr.12:31069845 - 31107058 on Build GRCh38 Duplication DDX11 DDX11-AS1
dgv1417n100 Chr.12:31065753 - 31261539 on Build GRCh38 Gain OVOS2 DDX11 DDX11-AS1
esv2422447 Chr.12:30801877 - 31296881 on Build GRCh38 Duplication OVOS2 TSPAN11 FAM60A DDX11 LINC00941 DDX11-AS1
dgv17n64 Chr.12:31057180 - 31253973 on Build GRCh38 Gain OVOS2 DDX11 DDX11-AS1
nsv1045497 Chr.12:31060801 - 31305613 on Build GRCh38 Gain OVOS2 FAM60A DDX11 DDX11-AS1
esv2422308 Chr.12:31081854 - 31122236 on Build GRCh38 Duplication OVOS2 DDX11
dgv62e55 Chr.12:31072776 - 31261539 on Build GRCh38 Gain OVOS2 DDX11 DDX11-AS1
nsv832363 Chr.12:31057544 - 31227100 on Build GRCh38 Gain OVOS2 DDX11 DDX11-AS1
esv22975 Chr.12:31072087 - 31200891 on Build GRCh38 Gain OVOS2 DDX11 DDX11-AS1
dgv7e196 Chr.12:30919025 - 31193061 on Build GRCh38 Duplication OVOS2 TSPAN11 DDX11 DDX11-AS1
dgv192n27 Chr.12:31048783 - 31253973 on Build GRCh38 Gain OVOS2 DDX11 DDX11-AS1
nsv558030 Chr.12:30816741 - 31113353 on Build GRCh38 Gain TSPAN11 DDX11 DDX11-AS1
nsv428276 Chr.12:30972532 - 31338170 on Build GRCh38 Gain OVOS2 TSPAN11 FAM60A FLJ13224 DDX11 DDX11-AS1
dgv2464n54 Chr.12:31094164 - 31262057 on Build GRCh38 Gain OVOS2 DDX11
dgv26e203 Chr.12:31080036 - 31264457 on Build GRCh38 Gain OVOS2 DDX11

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
sister chromatid cohesion
viral process
DNA duplex unwinding
IRE1-mediated unfolded protein response
double-stranded DNA binding
single-stranded DNA binding
RNA binding
ATP-dependent DNA helicase activity
helicase activity
protein binding
ATP binding
DNA-dependent ATPase activity
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-548c9458fd-h97b8:80/100.66.78.50:80.
git-commit: b50e517baec6e51dce58f17cb59caa961a96861c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.0-Offline