This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04464203_cn
See other LOC100130950 CNV Assays ›
Gene Symbol
LOC100130950
Assay Reference Genome
Location

Chr.17:5198529 on build GRCh38
Cytoband
17p13.2
Assay ID Hs04464203_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100130950

Gene Name:

uncharacterized LOC100130950

Gene Aliases:

-

Location:

Chr.17:5192084-5235636 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100130950 NR_034082.1
AK021878.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574273 Chr.17:4585589 - 5460332 on Build GRCh38 Gain DHX33 SLC25A11 SLC52A1 INCA1 TM4SF5 ALOX15 MINK1 SMTNL2 SPAG7 PLD2 ZNF594 LOC101927979 RABEP1 PFN1 MED11 RPAIN GLTPD2 LOC101559451 USP6 LOC102724009 ARRB2 CXCL16 PELP1 LOC101928000 ENO3 GP1BA C1QBP SCIMP LOC100130950 MIR6865 VMO1 CAMTA2 C17orf107 MIR6864 NUP88 ZFP3 KIF1C CHRNE LOC107984973 ZNF232 PSMB6 LOC400568 ZMYND15 RNF167
dgv3089n100 Chr.17:5187013 - 5327634 on Build GRCh38 Gain ZNF594 RABEP1 SCIMP LOC100130950

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0