This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04478892_cn
See other SERPINB5 CNV Assays ›
Gene Symbol
SERPINB5
Assay Reference Genome
Location

Chr.18:63481481 on build GRCh38
Cytoband
18q21.33
Assay ID Hs04478892_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

5268

Gene Name:

serpin family B member 5

Gene Aliases:

PI5, maspin

Location:

Chr.18:63476911-63505085 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SERPINB5 NM_002639.4 NP_002630.2
AK301667.1
AK312765.1
AL602439.1
BC020713.1 AAH20713.1
U04313.1 AAA18957.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1066552 Chr.18:63147299 - 64608176 on Build GRCh38 Gain SERPINB8 SERPINB10 SERPINB11 VPS4B LINC00305 SERPINB12 SERPINB5 SERPINB13 BCL2 KDSR HMSD LOC284294 SERPINB7 SERPINB3 LINC01538 SERPINB2 SERPINB4
nsv131597 Chr.18:63473298 - 63482700 on Build GRCh38 Deletion SERPINB5

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
movement of cell or subcellular component
negative regulation of endopeptidase activity
extracellular matrix organization
regulation of epithelial cell proliferation
prostate gland morphogenesis
serine-type endopeptidase inhibitor activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0