This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04532795_cn
See other MASP2 CNV Assays ›
Gene Symbol
MASP2
Assay Reference Genome
Location

Chr.1:11031604 on build GRCh38
Cytoband
1p36.22
Assay ID Hs04532795_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

10747

Gene Name:

mannan binding lectin serine peptidase 2

Gene Aliases:

MAP19, MASP-2, MASP1P1, sMAP

Location:

Chr.1:11026523-11047239 on Build GRCh38

Assay Gene Location:

Within Intron 9
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MASP2 NM_006610.3 NP_006601.2
AK290823.1
X98400.1 CAA67050.1
Y09926.1 CAA71059.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv832425 Chr.1:10843788 - 11062918 on Build GRCh38 Loss MASP2 TARDBP SRM C1orf127
nsv545407 Chr.1:11000018 - 11377969 on Build GRCh38 Gain ANGPTL7 UBIAD1 EXOSC10 LOC105376736 MTOR-AS1 MASP2 MTOR TARDBP SRM

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

complement activation, lectin pathway
proteolysis
complement activation
complement activation, classical pathway
complement component C4b binding
serine-type endopeptidase activity
calcium ion binding
protein binding
peptidase activity
calcium-dependent protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline