This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04540747_cn
See other SLC45A1 CNV Assays ›
Gene Symbol
SLC45A1
Assay Reference Genome
Location

Chr.1:8318739 on build GRCh38
Cytoband
1p36.23
Assay ID Hs04540747_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

50651

Gene Name:

solute carrier family 45 member 1

Gene Aliases:

DNB5

Location:

Chr.1:8318085-8344167 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC45A1 NM_001080397.2 NP_001073866.2
XM_011541530.1 XP_011539832.1
XM_011541531.1 XP_011539833.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1012977 Chr.1:8252522 - 8485210 on Build GRCh38 Loss SLC45A1 RERE LOC102724552
nsv1743 Chr.1:8317693 - 8351656 on Build GRCh38 Insertion SLC45A1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate transport
transmembrane transport
symporter activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline