This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04552421_cn
See other OMA1 CNV Assays ›
Gene Symbol
OMA1
Assay Reference Genome
Location

Chr.1:58500794 on build GRCh38
Cytoband
1p32.1
Assay ID Hs04552421_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

115209

Gene Name:

OMA1 zinc metallopeptidase

Gene Aliases:

2010001O09Rik, DAB1, MPRP-1, YKR087C, ZMPOMA1, peptidase

Location:

Chr.1:58480719-58546799 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
OMA1 NM_145243.4 NP_660286.1
AB048348.1 BAC79381.1
AK091101.1 BAC03583.1
BC012915.1 AAH12915.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv829948 Chr.1:58338009 - 58505708 on Build GRCh38 Gain OMA1
nsv1003930 Chr.1:58464772 - 58691366 on Build GRCh38 Gain OMA1 TACSTD2 MYSM1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

metalloprotease protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

diet induced thermogenesis
glucose metabolic process
misfolded or incompletely synthesized protein catabolic process
lipid metabolic process
response to stress
negative regulation of mitochondrial fusion
mitochondrial protein processing
cristae formation
energy homeostasis
metalloendopeptidase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0