This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04571308_cn
See other COMMD1 CNV Assays ›
Gene Symbol
COMMD1
Assay Reference Genome
Location

Chr.2:61890931 on build GRCh38
Cytoband
2p15
Assay ID Hs04571308_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

150684

Gene Name:

copper metabolism domain containing 1

Gene Aliases:

C2orf5, MURR1

Location:

Chr.2:61888723-62136070 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
COMMD1 NM_001321781.1 NP_001308710.1
NM_001321782.1 NP_001308711.1
DT217730.1
HY039942.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3591027 Chr.2:61889450 - 61893138 on Build GRCh38 Loss COMMD1
nsv582123 Chr.2:61719360 - 62324337 on Build GRCh38 Gain MIR5192 COMMD1 B3GNT2 CCT4 FAM161A
nsv582122 Chr.2:61692252 - 62020316 on Build GRCh38 Gain COMMD1 CCT4 FAM161A
nsv470467 Chr.2:61786803 - 62020316 on Build GRCh38 Gain COMMD1 CCT4 FAM161A
nsv582118 Chr.2:61431550 - 62075525 on Build GRCh38 Gain COMMD1 XPO1 CCT4 USP34 FAM161A
esv34000 Chr.2:61672382 - 62157212 on Build GRCh38 Loss COMMD1 CCT4 FAM161A
nsv1009446 Chr.2:61872503 - 61972765 on Build GRCh38 Gain COMMD1 CCT4

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
Golgi to plasma membrane transport
protein transport
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
regulation of proteasomal ubiquitin-dependent protein catabolic process
plasma membrane to endosome transport
copper ion homeostasis
negative regulation of sodium ion transmembrane transport
negative regulation of protein localization to cell surface
copper ion binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-3,4-bisphosphate binding
phosphatidic acid binding
phosphatidylinositol-3,5-bisphosphate binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0