This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04621006_cn
See other EIF4E2 CNV Assays ›
Gene Symbol
EIF4E2
Assay Reference Genome
Location

Chr.2:232561058 on build GRCh38
Cytoband
2q37.1
Assay ID Hs04621006_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9470

Gene Name:

eukaryotic translation initiation factor 4E family member 2

Gene Aliases:

4E-LP, 4EHP, EIF4EL3, IF4e

Location:

Chr.2:232550587-232583645 on Build GRCh38

Assay Gene Location:

Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
EIF4E2 NM_001276336.1 NP_001263265.1
NM_001276337.1 NP_001263266.1
NM_001282958.1 NP_001269887.1
NM_004846.3 NP_004837.1
XM_005246975.3 XP_005247032.1
XM_011512206.2 XP_011510508.1
XM_017005377.1 XP_016860866.1
AB209519.1 BAD92756.1
AF038957.1 AAC39871.1
AF047695.1 AAC18565.1
AF068117.1 AAC19374.1
AK022518.1
AK303802.1
BC005392.1 AAH05392.1
BC005874.2 AAH05874.1
BC021226.2 AAH21226.1
BC021690.1 AAH21690.1
BG913475.1
CD684451.1
DA408433.1
DA992544.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv3212 Chr.2:232526220 - 232564112 on Build GRCh38 Deletion EIF4E2 TIGD1 CHRND CHRNG MIR5001

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
translational initiation
negative regulation of translation
RNA cap binding
translation initiation factor activity
protein binding
translation factor activity, RNA binding
ubiquitin protein ligase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline