This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04668240_cn
See other SERPINE2 CNV Assays ›
Gene Symbol
SERPINE2
Assay Reference Genome
Location

Chr.2:223985592 on build GRCh38
Cytoband
2q36.1
Assay ID Hs04668240_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

5270

Gene Name:

serpin family E member 2

Gene Aliases:

GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI

Location:

Chr.2:223975048-224039319 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SERPINE2 NM_001136528.1 NP_001130000.1
NM_001136530.1 NP_001130002.1
NM_006216.3 NP_006207.1
NR_073116.1
XM_005246641.2 XP_005246698.1
XM_017004329.1 XP_016859818.1
XM_017004330.1 XP_016859819.1
XM_017004331.1 XP_016859820.1
XM_017004332.1 XP_016859821.1
A03911.1 CAA00310.1
AK225823.1
AK295564.1
AK297588.1
AK312499.1
BC015663.2 AAH15663.1
BC042628.1 AAH42628.1
M17783.1 AAA35883.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

embryo implantation
blood coagulation
negative regulation of cell proliferation
response to radiation
negative regulation of plasminogen activation
negative regulation of sodium ion transport
negative regulation of endopeptidase activity
negative regulation of protein processing
positive regulation of neuron projection development
negative regulation of phosphatidylinositol 3-kinase signaling
cerebellar granular layer morphogenesis
negative regulation of blood coagulation
negative regulation of cell growth
regulation of cell migration
secretion by cell
secretory granule organization
negative regulation of protein catabolic process
mating plug formation
negative regulation of proteolysis
negative regulation of smoothened signaling pathway
regulation of timing of cell differentiation
positive regulation of astrocyte differentiation
detection of mechanical stimulus involved in sensory perception
regulation of synaptic transmission, glutamatergic
long-term synaptic potentiation
innervation
seminal vesicle epithelium development
dense core granule biogenesis
negative regulation of platelet aggregation
serine-type endopeptidase inhibitor activity
receptor binding
protein binding
glycosaminoglycan binding
heparin binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline