This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04669026_cn
See other HSPE1-MOB4 CNV Assays ›
Gene Symbol
HSPE1-MOB4
Assay Reference Genome
Location

Chr.2:197512118 on build GRCh38
Cytoband
2q33.1
Assay ID Hs04669026_cn
Size
Availability Made To Order
Catalog # 4400291
Price 475.00
Your Price
Online offer: 441.65
441.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100529241

Gene Name:

HSPE1-MOB4 readthrough

Gene Aliases:

HSPE1-PHOCN

Location:

Chr.2:197499997-197553699 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HSPE1-MOB4 NM_001202485.1 NP_001189414.1
BJ992310.1
CR990871.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1014881 Chr.2:197432432 - 197668882 on Build GRCh38 Gain HSPE1-MOB4 RFTN2 HSPD1 SNORA105B SF3B1 HSPE1 COQ10B MOB4

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

kinase activator

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0